PloS one
Activation of p38 Mitogen-Activated Protein Kinase in Gaucher's Disease.
PMID 26312487
Abstract
Gaucher's disease is caused by defects in acid β-glucosidase 1 (GBA1) and has been also proposed as an inflammatory disease. GBA1 cleaves glucosylceramide to form ceramide, an established bioactive lipid, and defects in GBA1 lead to aberrant accumulation in glucosylceramide and insufficient formation of ceramide. We investigated if the pro-inflammatory kinase p38 is activated in Gaucher's disease, since ceramide has been proposed to suppress p38 activation. Three Gaucher's disease mouse models were employed, and p38 was found to be activated in lung and liver tissues of all Gaucher's disease mice. Most interestingly, neuronopathic Gaucher's disease type mice, but not non-neuronopathic ones, displayed significant activation of p38 and up-regulation of p38-inducible proinflammatory cytokines in brain tissues. In addition, all type of Gaucher's disease mice also showed increases in serum IL-6. As cellular signalling is believed to represent an in vivo inflammatory phenotype in Gaucher's disease, activation of p38 and possibly its-associated formation of proinflammatory cytokines were assessed in fibroblasts established from neuronopathic Gaucher's disease mice. In mouse Gaucher's disease cells, p38 activation and IL-6 formation by TNF-α treatment were enhanced as compared to those of wild type. Furthermore, human fibroblasts from Gaucher's disease patients also displayed increases in p38 activation and IL-6 formation as comparison to healthy counterpart. These results raise the potential that proinflammatory responses such as p38 activation and IL-6 formation are augmented in Gaucher's disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7556 Anti-MAP Kinase 2 (ERK-2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution, ~1 mg/mL
SAB4300499
Anti-MAPK14 (Ab-182) antibody produced in rabbit, affinity isolated antibody
SAB1405665
Anti-MAPK14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV06004
Anti-MAPK14 antibody produced in rabbit, affinity isolated antibody
M0800
Anti-p38 MAP Kinase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4500491
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500492
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500493
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500490
Anti-p38 MAPK, C-Terminal antibody produced in rabbit, affinity isolated antibody
GW21175B Anti-RK antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4502906
Anti-Synaptophysin antibody produced in rabbit, affinity isolated antibody
SAB4200544
Anti-Synaptophysin antibody, Mouse monoclonal, clone SVP-38, purified from hybridoma cell culture
HPA002858
Anti-SYP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102352
Anti-SYP antibody produced in rabbit, affinity isolated antibody
RAB0306
Human IL-6 ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0307
Human IL-6 ELISA Kit, for cell and tissue lysates
RAB0313
Human IL-6 sR ELISA Kit, for serum, plasma, cell culture supernatant and urine
M7431 Monoclonal Anti-MAP Kinase 2 (ERK-2) antibody produced in mouse, clone 1B3B9, purified immunoglobulin, buffered aqueous solution
SAB1403701
Monoclonal Anti-MAPK14, (C-terminal) antibody produced in mouse, clone 3D5, ascites fluid
S5768
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
WH0006855M1
Monoclonal Anti-SYP antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution
RAB0308
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0309
Mouse IL-6 ELISA Kit, for cell and tissue lysates
RAB0314 Mouse IL-6 R ELISA Kit, for serum, plasma and cell culture supernatant
RAB0311
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
RAB0312
Rat Interleukin-6 ELISA Kit, for for cell lysate and tissue lysate