EMAIL THIS PAGE TO A FRIEND
Cell death & disease
CD40 ligand induces RIP1-dependent, necroptosis-like cell death in low-grade serous but not serous borderline ovarian tumor cells.
PMID 26313915
Abstract
Ovarian high-grade serous carcinomas (HGSCs) and invasive low-grade serous carcinomas (LGSCs) are considered to be distinct entities. In particular, LGSCs are thought to arise from non-invasive serous borderline ovarian tumors (SBOTs) and show poor responsiveness to conventional chemotherapy. The pro-apoptotic effects of CD40 ligand (CD40L) have been demonstrated in HGSC, though the underlying mechanisms are not fully understood. Conversely, the therapeutic potential of the CD40L-CD40 system has yet to be evaluated in LGSC. We now show that CD40 protein is focally expressed on tumor cells in two of five primary LGSCs compared with no expression in eight primary SBOTs. Treatment with CD40L or agonistic CD40 antibody decreased the viability of LGSC-derived MPSC1 and VOA1312 cells, but not SBOT3.1 cells. Small interfering RNA (siRNA) targeting CD40 was used to show that it is required for these reductions in cell viability. CD40L treatment increased cleaved caspase-3 levels in MPSC1 cells though, surprisingly, neither pan-caspase inhibitor nor caspase-3 siRNA reversed or even attenuated CD40L-induced cell death. In addition, CD40-induced cell death was not affected by knockdown of the mitochondrial proteins apoptosis-inducing factor (AIF) and endonuclease G (EndoG). Interestingly, CD40L-induced cell death was blocked by necrostatin-1, an inhibitor of receptor-interacting protein 1 (RIP1), and attenuated by inhibitors of RIP3 (GSK'872) or MLKL (mixed lineage kinase domain-like; necrosulfonamide). Our results indicate that the upregulation of CD40 may be relatively common in LGSC and that CD40 activation induces RIP1-dependent, necroptosis-like cell death in LGSC cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV41929
Anti-CD40 antibody produced in rabbit, affinity isolated antibody
HPA031567
Anti-CD40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031568
Anti-CD40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103449
Anti-CD40, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4503674
Anti-CD40, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA046651
Anti-RALBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410141
Anti-RALBP1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2105823
Anti-RALBP1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
GW21168 Anti-RIPK1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA015257
Anti-RIPK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV02038
Anti-RIPK1 antibody produced in rabbit, affinity isolated antibody
SAB3500420
Anti-RIPK1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1401396
Anti-UQCRFS1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401397
Anti-UQCRFS1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA041863
Anti-UQCRFS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105078
Anti-UQCRFS1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
WH0000958M1
Monoclonal Anti-CD40 antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
AMAB90905
Monoclonal Anti-CD40 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1673, purified immunoglobulin, buffered aqueous glycerol solution
WH0010928M2
Monoclonal Anti-RALBP1 antibody produced in mouse, clone 2A1, purified immunoglobulin, buffered aqueous solution
SAB1404880 Monoclonal Anti-RALBP1 antibody produced in mouse, clone 1C2-A10, purified immunoglobulin, buffered aqueous solution
SAB4100176
Monoclonal Anti-RALBP1 antibody produced in mouse, clone RALBP1-A192, culture supernatant