PloS one
Pharmacokinetics, Tissue Distribution, and Anti-Lipogenic/Adipogenic Effects of Allyl-Isothiocyanate Metabolites.
PMID 26317351
Abstract
Allyl-isothiocyanate (AITC) is an organosulfur phytochemical found in abundance in common cruciferous vegetables such as mustard, wasabi, and cabbage. Although AITC is metabolized primarily through the mercapturic acid pathway, its exact pharmacokinetics remains undefined and the biological function of AITC metabolites is still largely unknown. In this study, we evaluated the inhibitory effects of AITC metabolites on lipid accumulation in vitro and elucidated the pharmacokinetics and tissue distribution of AITC metabolites in rats. We found that AITC metabolites generally conjugate with glutathione (GSH) or N-acetylcysteine (NAC) and are distributed in most organs and tissues. Pharmacokinetic analysis showed a rapid uptake and complete metabolism of AITC following oral administration to rats. Although AITC has been reported to exhibit anti-tumor activity in bladder cancer, the potential bioactivity of its metabolites has not been explored. We found that GSH-AITC and NAC-AITC effectively inhibit adipogenic differentiation of 3T3-L1 preadipocytes and suppress expression of PPAR-γ, C/EBPα, and FAS, which are up-regulated during adipogenesis. GSH-AITC and NAC-AITC also suppressed oleic acid-induced lipid accumulation and lipogenesis in hepatocytes. Our findings suggest that AITC is almost completely metabolized in the liver and rapidly excreted in urine through the mercapturic acid pathway following administration in rats. AITC metabolites may exert anti-obesity effects through suppression of adipogenesis or lipogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7018
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
C10H14N4O2
I5879
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
C10H14N4O2
D1756
Dexamethasone, ≥98% (HPLC), powder
C22H29FO5
D4902
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
C22H29FO5
D8893
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
C22H29FO5
D9184
Dexamethasone, meets USP testing specifications
C22H29FO5
O1008
Oleic acid, ≥99% (GC)
C18H34O2
O1383
Oleic acid, suitable for cell culture, BioReagent
C18H34O2
364525
Oleic acid, technical grade, 90%
C18H34O2
27728
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
C18H34O2
W281506
Oleic acid, natural, FCC
C18H34O2
P5811
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
H3O4P
466123
Phosphoric acid, crystalline, ≥99.999% trace metals basis
H3O4P
452289
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
H3O4P
345245
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
H3O4P
79617
Phosphoric acid, BioUltra, ≥85% (T)
H3O4P
40278
Phosphoric acid, semiconductor grade MOS PURANAL (Honeywell 17938), ≥85%
H3O4P
40266
Phosphoric acid, semiconductor grade PURANAL (Honeywell 17861), ≥85%
H3O4P
612162
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
H3O4P
699608
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
H3O4P
698822
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
H3O4P
W290017
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
H3O4P
609781
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
H316O4P
S7920
Sodium fluoride, BioXtra, ≥99%
FNa
S6776
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
FNa
215309
Sodium fluoride, 99.99% trace metals basis
FNa
71519
Sodium fluoride, BioUltra, ≥99.0% (F)
FNa
919
Sodium fluoride solution
FNa
792500
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
C3H3NaO3
80443
Sodium pyruvate, PharmaGrade, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C3H3NaO3