EMAIL THIS PAGE TO A FRIEND
Inflammation
Role of Suppressor of Cytokine Signaling 3 in the Immune Modulation of Mesenchymal Stromal Cells.
PMID 26318865
Abstract
The underlying mechanisms of mesenchymal stromal cells (MSCs) on immune modulation to treat allergic diseases remain unclear. Here, we showed that the suppressor of cytokine signaling 3 (SOCS3) is an important immune modulator expressed by MSCs, which is significantly increased by interferon-γ (IFN-γ). In addition, we observed that SOCS3 is a crucial mediator of the anti-proliferative and functional effects of MSCs on T cells and B cells. The immune modulation of MSCs through SOCS3 is mediated by cell-cell contacts. Moreover, SOCS3 could serve as an indicator to predict the potential immune modulatory of MSCs derived from different donors. Furthermore, treatment with anti-SOCS3 Ab significantly decreased ovalbumin-specific antibodies and neutrophil infiltration in ovalbumin-induced allergic rhinitis (AR) mice. Our results suggest that SOCS3 serves as an immune modulator interfering with T cells and B cells, and SOCS3 may act as a predictive marker for immune modulatory of MSCs.
Related Materials