EMAIL THIS PAGE TO A FRIEND
Journal of food protection
Investigation into the Efficacy of Bdellovibrio bacteriovorus as a Novel Preharvest Intervention To Control Escherichia coli O157:H7 and Salmonella in Cattle Using an In Vitro Model.
PMID 26319730
Abstract
Cattle are an important reservoir for the foodborne pathogens Salmonella and Escherichia coli O157:H7; they frequently harbor these microorganisms in their digestive tracts and shed them in their feces. Thus, there is potential for contamination of cattle hides and, subsequently, carcasses. Interventions aimed at reducing or eliminating pathogen shedding preharvest will also reduce the likelihood of beef product contamination by these pathogens. Therefore, this study used an in vitro model to evaluate Bdellovibrio bacteriovorus, a gram-negative microorganism that preys upon other gram-negative microorganisms, as a preharvest intervention to control Salmonella and E. coli O157:H7. Rumen fluid and feces were inoculated with pansusceptible or antimicrobial-resistant strains of one pathogen. Control samples were treated with HEPES buffer, whereas experimental samples were exposed to HEPES buffer plus B. bacteriovorus. Salmonella and E. coli O157:H7 populations were quantified at 0, 24, 48, and 72 h. The most-probable-number (MPN) technique, followed by streaking onto xylose lysine Tergitol 4 agar, was used to determine Salmonella populations, whereas spread plating onto sorbitol MacConkey agar supplemented with cefixime and tellurite was employed to enumerate E. coli O157:H7. B. bacteriovorus reduced pansusceptible Salmonella in cattle feces by 2.02 Log MPN/g (P = 0.0005) and antimicrobial-resistant Salmonella by 3.79 (P < 0.0001) and 2.24 (P = 0.0013) Log MPN/g after 24 and 48 h, respectively, in comparison to control samples. Significant reductions were not observed for E. coli O157:H7 in rumen or feces. These data suggest that further investigation into B. bacteriovorus efficacy as a preharvest intervention to control Salmonella in cattle is warranted.