EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Ultra high performance liquid chromatography coupled to tandem mass spectrometry determination of lipid peroxidation biomarkers in newborn serum samples.
PMID 26320656
Abstract
Byproducts of arachidonic (AA) and docosahexaenoic acid (DHA) oxidation are highly relevant for the study of free radical associated conditions in the perinatal period. Plasma metabolites can provide the clinician with a snapshot of the oxidant status of patients before and after specific clinical interventions (e.g.: supplementation with oxygen). We describe a new andreliable ultra-performance liquid mass spectrometry method to determine F2-isoprostanes and other byproducts (isoprostanes, isofurans, neuroprostanes, neurofurans) in newborn serum samples. Cord blood samples were obtained from severely depressed newborn infants (Apgar score 1 min < 3; arterial cord pH < 7.00), and aliquoted for serum determination and stored at -80 °C. A UHPLC-MS/MS method was employed. It has a series of technical advantages: simple sample treatment; reduced sample volume (100 μL) which is essential for preterm neonates with low circulating blood volume, high throughput of sample analysis (96 samples in less than 24 h) and high selectivity for different isoprostanes isomers. Excellent sensitivity was achieved within limits of detection between 0.06 and 4.2 nmol L(-1), which renders this method suitable to monitoranalyte concentration in newborn samples. The method's precision was satisfactory; with coefficients of variation around 5-12% (intra-day) and 7-17% (inter-day). The reliability of the described method was assessed by analysis of spiked serum samples obtaining recoveries between 70% and 120%. The proposed method has rendered suitable for serum determination for newborn babies at risk of oxygen free radical associated conditions.