EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Determination of phthalate esters in vegetable oils using direct immersion solid-phase microextraction and fast gas chromatography coupled with triple quadrupole mass spectrometry.
PMID 26320808
Abstract
Phthalates are a group of synthetic compounds mainly used as plasticizers, which have been classified as endocrine-disrupting chemicals and potential human-cancer causing agents. They can be found in high amounts in foods, deriving mainly from plastic packaging. The analytical determination of these compounds is very challenging since they are ubiquitous. Therefore, minimization of sample manipulation is highly desirable. The present work exploited the application of a solid-phase microextraction method for the analysis of phthalates in vegetable oil. A preliminary comparison between a polydimethylsiloxane (PDMS) and a Carbopack Z/PDMS fiber was carried out both in the headspace and direct immersion extraction modes. Before immersing the fiber, a rapid liquid-liquid extraction was performed using acetonitrile to remove the bulk of triglycerides. PDMS in the direct immersion mode showed the best performance. The method was fully validated obtaining a good linearity with a coefficient of correlation of over 0.9960 for all compounds, repeatability and accuracy values generally better than 10%, and very good limit of quantification values.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00561
Acetone, for luminescence, ≥99.5% (GC)
C3H6O
40289
Acetone, semiconductor grade VLSI PURANAL (Honeywell 17617)
C3H6O
W332607
Acetone, ≥99%, meets FCC analytical specifications
C3H6O
W332615
Acetone, natural, ≥97%
C3H6O
01-0520
Acetone, for HPLC
C3H6O
01-0494
Acetone, for residue analysis, suitable for 5000 per JIS
C3H6O
01-0475
Acetone, suitable for determination of dioxins
C3H6O
686530
Acetone solution, contains 20.0 % (v/v) acetonitrile, for HPLC
C3H6O
753939
DBP, 98% (HPLC)
C64H36
274623
Dess-Martin periodinane, 97%
C13H13IO8
D2270
Dibutyl phthalate
C16H22O4
152439
Dibutyl phthalate, ReagentPlus®, ≥99%
C16H22O4
524980
Dibutyl phthalate, 99%
C16H22O4
306150
Dicyclohexyl phthalate, 99%
C20H26O4
524972
Diethyl phthalate, 99.5%
C12H14O4
W512206
Diethyl phthalate, ≥99%
C12H14O4
40718
Diethyl pyrocarbonate, 99% (NT)
C6H10O5
D5758
Diethyl pyrocarbonate, ≥97% (NMR)
C6H10O5
159220
Diethyl pyrocarbonate, 97%
C6H10O5
152641
Diisobutyl phthalate, 99%
C16H22O4
376663
Diisononyl phthalate, ester content ≥99 % (mixture of C9 isomers), technical grade
C26H42O4
W508500
Dimethyl phthalate, ≥99%
C10H10O4
525081
Dimethyl phthalate, ≥99%
C10H10O4
07-4770
Dimethyl phthalate, SAJ special grade, ≥99.0%
C10H10O4
290602
Dipropyl phthalate, 98%
C14H18O4
71649
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
HNa2O4P · 12H2O