EMAIL THIS PAGE TO A FRIEND
Cancer letters
ABL SH3 mutant inhibits BCR-ABL activity and increases imatinib sensitivity by targeting RIN1 protein in CML cell.
PMID 26321052
Abstract
SH3 domain plays an important role in maintaining autoinhibition of BCR-ABL protein. RIN1 interacts with BCR-ABL SH3 domain via PxxP motifs to promote autophosphorylation as well as activation of BCR-ABL tyrosine kinase, suggesting using exogenous SH3 domain which blocks the interaction of BCR-ABL and RIN1 could be an adjunct therapy for CML. Here, we reported a novel p-BCR-ABL inhibitor, designed as ABL SH3 mutant, and identified its effects on inhibiting the tyrosine kinase activity of BCR-ABL without or with imatinib (IM) in vitro and in vivo. Our results demonstrated that ABL SH3 mutant T79Y markedly repressed the expression of BCR-ABL signaling pathways in IM-resistant cell lines KCL22 and K562/G01 as well as IM-sensitive cell line K562. Moreover, combination of T79Y with IM considerably decreased the proliferation of leukemia cells in vivo. Inhibition of BCR-ABL and RIN1 interaction using exogenous modified BCR-ABL SH3 domain provides a feasible and alternative option of small molecule inhibitors for CML treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501043
Anti-ABL antibody produced in rabbit, affinity isolated antibody
HPA027251
Anti-ABL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA027280
Anti-ABL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA028409
Anti-ABL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501046
Anti-ABL1 antibody produced in rabbit, affinity isolated antibody
SAB4501047
Anti-ABL1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2500274
Anti-CRKL antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405659
Anti-CRKL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001100
Anti-CRKL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV01043
Anti-CRKL antibody produced in rabbit, affinity isolated antibody
SAB4500471
Anti-CRKL antibody produced in rabbit, affinity isolated antibody
SAB1405763
Anti-EPHB1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4500732
Anti-EPHB1 antibody produced in rabbit, affinity isolated antibody
SAB4500775
Anti-EPHB1 antibody produced in rabbit, affinity isolated antibody
SAB4500776
Anti-EPHB1 antibody produced in rabbit, affinity isolated antibody
SAB1412026
ANTI-EPHB2 antibody produced in mouse, clone 1F10, ascites fluid, buffered aqueous solution
SAB4500777
Anti-EPHB2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300037
Anti-phospho-STAT5A (pSer780) antibody produced in rabbit, affinity isolated antibody
SAB4504543
Anti-phospho-STAT5A (pSer780) antibody produced in rabbit, affinity isolated antibody
SAB4300036
Anti-phospho-STAT5A (pTyr694) antibody produced in rabbit, affinity isolated antibody
SAB4504544
Anti-phospho-STAT5A (pTyr694) antibody produced in rabbit, affinity isolated antibody
SAB1406856
Anti-RIN1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA035491
Anti-RIN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411683
Anti-RIN1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4503678
Anti-RIN1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300329
Anti-STAT5A (Ab-694) antibody produced in rabbit, affinity isolated antibody
SAB4300330
Anti-STAT5A (Ab-780) antibody produced in rabbit, affinity isolated antibody
SAB1406488
Anti-STAT5A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2102318
Anti-STAT5A antibody produced in rabbit, affinity isolated antibody
HPA027873
Anti-STAT5A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution