EMAIL THIS PAGE TO A FRIEND
Gut pathogens
High dietary zinc supplementation increases the occurrence of tetracycline and sulfonamide resistance genes in the intestine of weaned pigs.
PMID 26322131
Abstract
Dietary zinc oxide is used in pig nutrition to combat post weaning diarrhoea. Recent data suggests that high doses (2.5xa0g/kg feed) increase the bacterial antibiotic resistance development in weaned pigs. Therefore, the aim of this study was to investigate the development of enterobacterial antibiotic resistance genes in the intestinal tract of weaned pigs. Weaned pigs were fed diets for 4xa0weeks containing 57 (low), 164 (intermediate) or 2425 (high) mgxa0kg(-1) analytical grade ZnO. DNA extracts from stomach, mid-jejunum, terminal ileum and colon ascendens were amplified by qPCR assays to quantify copy numbers for the tetracycline (tetA) and sulfonamide (sul1) resistance genes in Gram-negative bacteria. Overall, the combined data (nxa0=xa0336) showed that copy numbers for tetracycline and sulfonamide resistance genes were significantly increased in the high zinc treatment compared to the low (tetA: p valuexa0