EMAIL THIS PAGE TO A FRIEND
Nature chemical biology
Extracellular 4'-phosphopantetheine is a source for intracellular coenzyme A synthesis.
PMID 26322826
Abstract
The metabolic cofactor coenzyme A (CoA) gained renewed attention because of its roles in neurodegeneration, protein acetylation, autophagy and signal transduction. The long-standing dogma is that eukaryotic cells obtain CoA exclusively via the uptake of extracellular precursors, especially vitamin B5, which is intracellularly converted through five conserved enzymatic reactions into CoA. This study demonstrates an alternative mechanism that allows cells and organisms to adjust intracellular CoA levels by using exogenous CoA. Here CoA was hydrolyzed extracellularly by ectonucleotide pyrophosphatases to 4'-phosphopantetheine, a biologically stable molecule able to translocate through membranes via passive diffusion. Inside the cell, 4'-phosphopantetheine was enzymatically converted back to CoA by the bifunctional enzyme CoA synthase. Phenotypes induced by intracellular CoA deprivation were reversed when exogenous CoA was provided. Our findings answer long-standing questions in fundamental cell biology and have major implications for the understanding of CoA-related diseases and therapies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4780
Coenzyme A sodium salt hydrate, suitable for cell culture, BioReagent
C21H36N7O16P3S · xNa+ · yH2O