EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Degradation of HK2 by chaperone-mediated autophagy promotes metabolic catastrophe and cell death.
PMID 26323688
Abstract
Hexokinase II (HK2), a key enzyme involved in glucose metabolism, is regulated by growth factor signaling and is required for initiation and maintenance of tumors. Here we show that metabolic stress triggered by perturbation of receptor tyrosine kinase FLT3 in non-acute myeloid leukemia cells sensitizes cancer cells to autophagy inhibition and leads to excessive activation of chaperone-mediated autophagy (CMA). Our data demonstrate that FLT3 is an important sensor of cellular nutritional state and elucidate the role and molecular mechanism of CMA in metabolic regulation and mediating cancer cell death. Importantly, our proteome analysis revealed that HK2 is a CMA substrate and that its degradation by CMA is regulated by glucose availability. We reveal a new mechanism by which excessive activation of CMA may be exploited pharmacologically to eliminate cancer cells by inhibiting both FLT3 and autophagy. Our study delineates a novel pharmacological strategy to promote the degradation of HK2 in cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8203
Anti-α-Tubulin antibody, Mouse monoclonal, clone AA13, purified from hybridoma cell culture
SAB4500797
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500799
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500800
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500802
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500803
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500796
Anti-AKT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500798
Anti-AKT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500004
Anti-AKT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300335
Anti-AKT1 (Ab-308) antibody produced in rabbit, affinity isolated antibody
SAB4300575
Anti-AKT1 (Ab-450) antibody produced in rabbit, affinity isolated antibody
SAB4300334
Anti-AKT1 (Ab-473) antibody produced in rabbit, affinity isolated antibody
GW22757 Anti-AKT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22775 Anti-AKT1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501934
Anti-AKT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3500216
Anti-Akt1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA002891
Anti-AKT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV06008 Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
SAB4500007
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
AV36578
Anti-ANXA3 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV36579
Anti-ANXA3 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1405458
Anti-ANXA3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA013398
Anti-ANXA3 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA013431
Anti-ANXA3 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PRS3611
Anti-Beclin-1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3613
Anti-Beclin-1 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2103299
Anti-BECN1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4503294
Anti-CASP3 (p17, Cleaved-Asp175) antibody produced in rabbit, affinity isolated antibody
SAB2501669
Anti-CASP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV00021
Anti-CASP3 antibody produced in rabbit, IgG fraction of antiserum