EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Usp16 regulates kinetochore localization of Plk1 to promote proper chromosome alignment in mitosis.
PMID 26323689
Abstract
During the G2 to M phase transition, a portion of mitotic regulator Plk1 localizes to the kinetochores and regulates the initiation of kinetochore-microtubule attachments for proper chromosome alignment. Once kinetochore-microtubule attachment is achieved, this portion of Plk1 is removed from the kinetochores as a result of ubiquitination. However, the crucial molecular mechanism that promotes the localization and the maintenance of Plk1 on the kinetochores until metaphase is still unclear. We report that ubiquitin-specific peptidase 16 (Usp16) plays a key role during this process. Usp16 deubiquitinates Plk1, resulting in an enhanced interaction with kinetochore-localized proteins such as BubR1, and thereby retains Plk1 on the kinetochores to promote proper chromosome alignment in early mitosis. Down-regulation of Usp16 causes increased ubiquitination and decreased kinetochore localization of Plk1. Thus, our data unveil a unique mechanism by which Usp16 promotes the localization and maintenance of Plk1 on the kinetochores for proper chromosome alignment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9745
Anti-Cullin 3 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
C0871
Anti-Cullin 3 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2700710
Anti-KLHL22 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107560
Anti-KLHL22 antibody produced in rabbit, affinity isolated antibody
SAB1406265
Anti-PLK1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101829
Anti-PLK1 antibody produced in rabbit, affinity isolated antibody
HPA051638 Anti-PLK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA053229 Anti-PLK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502211
Anti-PLK1 antibody produced in rabbit, affinity isolated antibody
SAB1407022
Anti-USP16 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV45772
Anti-USP16 antibody produced in rabbit, IgG fraction of antiserum
HPA021140
Anti-USP16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306909
ANTI-USP21 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA028397
Anti-USP21 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1307083
ANTI-USP21(C-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1306999
ANTI-USP21(N-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
WH0008452M1
Monoclonal Anti-CUL3 antibody produced in mouse, clone 1A3, purified immunoglobulin, buffered aqueous solution
SAB4200180
Monoclonal Anti-Cullin 3 antibody produced in mouse, ~1.0 mg/mL, clone CUL3-9, purified immunoglobulin
SAB5300063
Monoclonal Anti-KLHL22 antibody produced in mouse, clone 7B10, ascites fluid
WH0005347M1
Monoclonal Anti-PLK1 antibody produced in mouse, clone 2G12, purified immunoglobulin, buffered aqueous solution
SAB1404220
Monoclonal Anti-PLK1 antibody produced in mouse, clone 4G11, purified immunoglobulin, buffered aqueous solution
WH0027005M3
Monoclonal Anti-USP21 antibody produced in mouse, clone 3D10, purified immunoglobulin, buffered aqueous solution
N9877
Nitrilotriacetic acid, Sigma Grade, ≥99%
C6H9NO6
72560
Nitrilotriacetic acid, ACS reagent, ≥99.0%
C6H9NO6
72559
Nitrilotriacetic acid, BioUltra, ≥99.0% (T)
C6H9NO6
PLA0073
Rabbit anti-PLK1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
T1895
Thymidine, powder, BioReagent, suitable for cell culture
C10H14N2O5
T9250
Thymidine, ≥99%
C10H14N2O5
89270
Thymidine, ≥99.0% (HPLC)
C10H14N2O5