EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Hsc70 chaperone activity underlies Trio GEF function in axon growth and guidance induced by netrin-1.
PMID 26323693
Abstract
During development, netrin-1 is both an attractive and repulsive axon guidance cue and mediates its attractive function through the receptor Deleted in Colorectal Cancer (DCC). The activation of Rho guanosine triphosphatases within the extending growth cone facilitates the dynamic reorganization of the cytoskeleton required to drive axon extension. The Rac1 guanine nucleotide exchange factor (GEF) Trio is essential for netrin-1-induced axon outgrowth and guidance. Here, we identify the molecular chaperone heat shock cognate protein 70 (Hsc70) as a novel Trio regulator. Hsc70 dynamically associated with the N-terminal region and Rac1 GEF domain of Trio. Whereas Hsc70 expression supported Trio-dependent Rac1 activation, adenosine triphosphatase-deficient Hsc70 (D10N) abrogated Trio Rac1 GEF activity and netrin-1-induced Rac1 activation. Hsc70 was required for netrin-1-mediated axon growth and attraction in vitro, whereas Hsc70 activity supported callosal projections and radial neuronal migration in the embryonic neocortex. These findings demonstrate that Hsc70 chaperone activity is required for Rac1 activation by Trio and this function underlies netrin-1/DCC-dependent axon outgrowth and guidance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300665
Anti-FAK (Ab-397) antibody produced in rabbit, affinity isolated antibody
SAB4502495
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502496
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502498
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502499
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502503
Anti-FAK antibody produced in rabbit, affinity isolated antibody
SAB4502497
Anti-FAK, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502500
Anti-FAK, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502501
Anti-FAK, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4502502
Anti-FAK, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2101096
Anti-HSPA4 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101097
Anti-HSPA4 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB1405971
Anti-HSPA4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA010023
Anti-HSPA4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410667
Anti-HSPA4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2701704
Anti-hspa8 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701706
Anti-hspa8 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701813
Anti-hspa8 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701964
Anti-HSPA8 antibody produced in rabbit
SAB2702050
Anti-HSPA8 antibody produced in rabbit
SAB2702054
Anti-HSPA8 antibody produced in rabbit
SAB2104919
Anti-HSPA8, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4503135
Anti-MAPK3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503424
Anti-MNK1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4504583
Anti-phospho-FAK (pSer843) antibody produced in rabbit, affinity isolated antibody
SAB4504180
Anti-phospho-FAK (pSer910) antibody produced in rabbit, affinity isolated antibody
F7926 Anti-phospho-FAK (pTyr397) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504181
Anti-phospho-FAK (pTyr397) antibody produced in rabbit, affinity isolated antibody
SAB4504403
Anti-phospho-FAK (pTyr397) antibody produced in rabbit, affinity isolated antibody
F8051 Anti-phospho-FAK (pTyr407) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution