EMAIL THIS PAGE TO A FRIEND
Biological & pharmaceutical bulletin
Anti-fatigue Effects of 20(S)-Protopanaxadiol and 20(S)-Protopanaxatriol in Mice.
PMID 26328499
Abstract
Ginseng (Panax ginseng C.A. MEYER, Araliaceae), which contains protopanaxadiol-type and protopanaxatriol-type ginsenosides, has been used for inflammation, fatigue, stress, and tumor in Asian countries. Orally administered ginsenosides are metabolized to their aglycones 20(S)-protopanaxadiol (PPD) and 20(S)-protopanaxatriol (PPT) by gut microbiota. However, their anti-fatigue effects have not been studied thoroughly. Therefore, we investigated the anti-fatigue activities of PPD and PPT in mice, using the weight-loaded swimming (WLS) and the rota-rod tests. Ginseng water extract (GW), ginseng saponin fraction (GWS) and ginseng polysaccharide fraction (GWP) at concentrations of 50 and 100 mg/kg and PPD and PPT at 5 and 10 mg/kg were orally administered to mice once daily for 5 d. GW, GWS, and PPT significantly increased the WLS time, however, GWP and PPD did not cause any significant change. PPT induced the most significant increase in WLS time. PPD (10 mg/kg) and PPT (5 and 10 mg/kg) inhibited the WLS-induced increase in corticosterone, lactate, lactate dehydrogenase (LDH), and creatinine levels as well as the reduction in glucose level. PPT increased the riding time in the rota-rod test, and also inhibited corticosterone, lactate, and creatinine levels. These findings suggest that the anti-fatigue effect of ginseng may be attributable to its saponins, particularly PPT, rather than to its polysaccharides.