EMAIL THIS PAGE TO A FRIEND
Ophthalmic research
Herpes virus entry mediator in human corneal epithelial cells modulates the production of inflammatory cytokines in response to HSV type 1 challenge.
PMID 26328884
Abstract
To investigate the role of herpes virus entry mediator (HVEM) in viral entry and inflammatory cytokine production in response to herpes simplex virus (HSV) type 1 challenge in human corneal epithelial cells. HVEM expression in human corneal epithelial cells was determined by immunofluorescence and flow cytometry. The HSV-1 virus expressing β-galactosidase was used to challenge corneal epithelial cells and viral entry assays were performed to ascertain HSV-1 entry into cells. Levels of cytokines TNF-α, IL-6, IFN-x03B3;, IL-12, and IL-18 and chemokines MIP-1α, MIP-1β and MIP-2 were detected in corneal epithelial cells treated with control or HVEM siRNA in response to HSV-1 challenge. Human corneal epithelial cells were positive for HVEM expression and showed high susceptibility to HSV-1 entry. Silencing of HVEM did not alter viral entry dramatically. However, levels of the cytokine IFN-x03B3; and chemokines MIP-1α and MIP-1β were measured to be higher in HVEM siRNA-treated cells than control after HSV-1 challenge. HVEM in human corneal epithelial cells may act to dampen the production of some cytokines and chemokines and thus it may modulate the innate immune response against HSV-1. This may provide a novel mechanism for the pathogenesis of HSV-1 infection in the cornea.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1127
2-Nitrophenyl β-D-galactopyranoside, ≥98% (enzymatic)
C12H15NO8