EMAIL THIS PAGE TO A FRIEND
Diagnostic pathology
On the representation of cells in bone marrow pathology by a scalar field: propagation through serial sections, co-localization and spatial interaction analysis.
PMID 26330285
Abstract
Immunohistochemical analysis of cellular interactions in the bone marrow in situ is demanding, due to its heterogeneous cellular composition, the poor delineation and overlap of functional compartments and highly complex immunophenotypes of several cell populations (e.g. regulatory T-cells) that require immunohistochemical marker sets for unambiguous characterization. To overcome these difficulties, we herein present an approach to describe objects (e.g. cells, bone trabeculae) by a scalar field that can be propagated through registered images of serial histological sections. The transformation of objects within images (e.g. cells) to a scalar field was performed by convolution of the object's centroids with differently formed radial basis function (e.g. for direct or indirect spatial interaction). On the basis of such a scalar field, a summation field described distributed objects within an image. After image registration i) colocalization analysis could be performed on basis scalar field, which is propagated through registered images, and - due to the shape of the field - were barely prone to matching errors and morphological changes by different cutting levels; ii) furthermore, depending on the field shape the colocalization measurements could also quantify spatial interaction (e.g. direct or paracrine cellular contact); ii) the field-overlap, which represents the spatial distance, of different objects (e.g. two cells) could be calculated by the histogram intersection. The description of objects (e.g. cells, cell clusters, bone trabeculae etc.) as a field offers several possibilities: First, co-localization of different markers (e.g. by immunohistochemical staining) in serial sections can be performed in an automatic, objective and quantifiable way. In contrast to multicolour staining (e.g. 10-colour immunofluorescence) the financial and technical requirements are fairly minor. Second, the approach allows searching for different types of spatial interactions (e.g. direct and indirect cellular interaction) between objects by taking field shape into account (e.g. thin vs. broad). Third, by describing spatially distributed groups of objects as summation field, it gives cluster definition that relies rather on the bare object distance than on the modelled spatial cellular interaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300361
Anti-ITGB3 (Ab-773) antibody produced in rabbit, affinity isolated antibody
SAB4300526
Anti-ITGB3 (Ab-785) antibody produced in rabbit, affinity isolated antibody
HPA027852
Anti-ITGB3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2502167
Anti-MPO antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA021147
Anti-MPO antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501128
Anti-MS4A1 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405580
Anti-MS4A1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA014341
Anti-MS4A1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA014391
Anti-MS4A1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4700114 Monoclonal Anti-CD20 antibody produced in mouse, clone 2H7, purified immunoglobulin, buffered aqueous solution
SAB4700119 Monoclonal Anti-CD20 antibody produced in mouse, clone MEM-269, purified immunoglobulin, buffered aqueous solution
SAB4700121
Monoclonal Anti-CD20 antibody produced in mouse, clone MEM-97, purified immunoglobulin, buffered aqueous solution
SAB1409321
Monoclonal Anti-MPO antibody produced in mouse, clone 3E11, purified immunoglobulin, buffered aqueous solution
WH0000931M1
Monoclonal Anti-MS4A1 antibody produced in mouse, clone 5C11, purified immunoglobulin, buffered aqueous solution
SAB1403646
Monoclonal Anti-MS4A1 antibody produced in mouse, clone 4F11, purified immunoglobulin, buffered aqueous solution