EMAIL THIS PAGE TO A FRIEND
Iranian journal of pharmaceutical research : IJPR
Enhanced Production of Insulin-like Growth Factor I Protein in Escherichia coli by Optimization of Five Key Factors.
PMID 26330880
Abstract
Human insulin-like growth factor I (hIGF-I) is a kind of growth factor with clinical significance in medicine. Up to now, E. coli expression system has been widely used as a host to produce rhIGF-1 with high yields. Batch cultures as non-continuous fermentations were carried out to overproduce rhIGF-I in E. coli. The major objective of this study is over- production of recombinant human insulin-like growth factor I (rhIGF-I) through a developed process by recruiting effective factors in order to achieve the most recombinant protein. In this study we investigated the effect of culture medium, induction temperature and amount of inducer on cell growth and IGF-1 production. Taguchi design of experiments (DOE) method was used as the statistical method. Analysis of experimental data showed that maximum production of rhIGF-I was occurred in 32y culture medium at 32 °C and 0.05 Mm IPTG. Under this condition, 0.694 g/L of rhIGF-I was produced as the inclusion bodies. Following optimization of these three factors, we have also optimized the amount of glucose and induction time in 5 liter top bench bioreactor. Full factorial design of experiment method was used for these two factors as the statistical method. 10 g/L and OD600=5 were selected as the optimum point of Glucose amount and induction time, respectively. Finally, we reached to a concentration of 1.26 g/L rhIGF-1 at optimum condition.