EMAIL THIS PAGE TO A FRIEND
Scientific reports
Establishment of immunoassay for detecting HPV16 E6 and E7 RNA.
PMID 26333509
Abstract
Cervical carcinoma is the most prevalent malignancy second only to breast cancer among women worldwide. Since more than 99% of cervical cancers are caused by human papilloma virus (HPV), measurement of HPV (HPV test) was commonly used in screening risk and/or early stage of cervical cancer as well as assessing the efficacies of the treatments that can decrease the incidence of cervical cancer. Many approaches that diagnose HPV infections have been developed, while most of them have distinct shortcomings. We here established a novel immunoassay method in which the pairs of unlabeled DNA probes firstly bind to HPV16 E6 and E7 RNAs to form the DNA-RNA hybrids, and the hybrids will subsequently be identified by S9.6 antibody. The sensitivity of this highly specific method can reach ~0.923 pg/mL and ~0.424 pg/mL of in vitro transcribed HPV16 E6 and E7 RNA, respectively, and reverse transcription and polymerase chain reaction (PCR) amplification were no longer needed. Thus, our immunoassay approaches can precisely reflect the actually viral load that is related to the course of HPV infection. In addition, it has also fast and low cost characteristic feature.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
860336
3,3′,5,5′-Tetramethylbenzidine, ≥99%
C16H20N2
T2885
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
C16H20N2
T5525
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
C16H20N2
87748
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
C16H20N2
A1293 Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A2179 Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A1682 Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B7151 Anti-Mouse IgG (Fab specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B0529 Anti-Mouse IgG (Fab specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F5262 Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F4018 Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F8771 Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A9917 Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A2304 Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A3682 Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
T6528
Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
T7782 Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701030 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701031 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701028 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Fluorescein antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701029 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701011 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Rhodamine antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701033 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Texas Red™ antibody produced in goat, affinity isolated antibody, lyophilized powder
A1418 Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A2429 Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A7434
Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
B7401 Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B9904 Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F5387 Anti-Mouse IgG (Fc specific)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
F4143 Anti-Mouse IgG (Fc specific)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution