EMAIL THIS PAGE TO A FRIEND
Scientific reports
FoxM1 Directs STAT3 Expression Essential for Human Endometrial Stromal Decidualization.
PMID 26334131
Abstract
Human endometrium decidualization, which involves endometrial stromal proliferation and differentiation, is a prerequisite for embryo implantation, thus successful pregnancy. The Forkhead Box M1 (FoxM1), previously known as HNF-3, HFH-11, MPP2, Win, and Trident, is a transcriptional factor that plays crucial roles in cell proliferation and cell cycle progression. However, the molecular mechanism of FoxM1 during human endometrial decidualization remains unexplored. In this study, we first found FoxM1 is dynamically expressed in human endometrium during menstrual cycle. Employing a human endometrial stromal cell (HESC) line, we then demonstrated that FoxM1 inhibition downregulates cyclin B1 expression, delaying G2/M phase transition during HESC proliferation. Additionally, loss of FoxM1 expression blocks the differentiation of HESCs in response to estrogen, progesterone, and dbcAMP. Applying chromatin immunoprecipitation (ChIP) technique and luciferase assay, we further approved that FoxM1 can transcriptionally active signal transducer and activator of transcription 3 (STAT3), ensuring normal HESC differentiation. Besides enriching our knowledge on molecular basis underlying stromal decidualization, these findings help to shed light on the potential molecular causes for the endometrial disorders in humans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H56800
2-Hydroxypyridine, 97%
C5H5NO
A9501
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
C10H12N5O6P
A3262
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
C10H12N5O6P · C4H11NO3
SAB4503294
Anti-CASP3 (p17, Cleaved-Asp175) antibody produced in rabbit, affinity isolated antibody
SAB2501669
Anti-CASP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA002643
Anti-CASP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV00021
Anti-CASP3 antibody produced in rabbit, IgG fraction of antiserum
SAB4503291
Anti-CASP3 antibody produced in rabbit, affinity isolated antibody
HPA001473
Anti-CASP9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503332
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503333
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503334
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
SAB4503335
Anti-CASP9 antibody produced in rabbit, affinity isolated antibody
C9598 Anti-Caspase 3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
C7729 Anti-Caspase 9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300683
Anti-Caspase 9 antibody produced in rabbit, affinity isolated antibody
SAB3500405
Anti-Caspase-9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV30161
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
SAB2100360
Anti-CCNB1 antibody produced in rabbit, affinity isolated antibody
HPA012024
Anti-CEBPG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV35645
Anti-CEBPG antibody produced in rabbit, affinity isolated antibody
SAB2105235
Anti-CEBPG antibody produced in rabbit, affinity isolated antibody
SAB4500123
Anti-CEBPG antibody produced in rabbit, affinity isolated antibody
HPA014101
Anti-CHRM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500244
Anti-CHRM1 antibody produced in rabbit, affinity isolated antibody
C8831
Anti-Cyclin B1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB4503501
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
SAB4503502
Anti-Cyclin B1 antibody produced in rabbit, affinity isolated antibody
SAB4500487
Anti-PARP (Cleaved-Asp214) antibody produced in rabbit, affinity isolated antibody
SAB4500488
Anti-PARP (Cleaved-Gly215) antibody produced in rabbit, affinity isolated antibody