EMAIL THIS PAGE TO A FRIEND
Applied biochemistry and biotechnology
Engineering of Alicyclobacillus hesperidum L-arabinose isomerase for improved catalytic activity and reduced pH optimum using random and site-directed mutagenesis.
PMID 26335445
Abstract
A mutation, D478N, was obtained by an error-prone polymerase chain reaction using the L-arabinose isomerase (L-AI) gene from Alicyclobacillus hesperidum URH17-3-68 as the template. The mutated isomerase showed higher activity for D-galactose isomerization. The mutation site obtained from random mutagenesis was then introduced as a single-site mutation using site-directed mutagenesis. Single-site variants, D478N, D478Q, D478A, D478K, and D478R, were constructed. The optimum temperatures were all higher than 60xa0°C. D478A, D478N, and D478Q retained more than 80xa0% of the maximum relative activity of the wild-type L-AI at 75xa0°C. With the exception of the D478A variant, all variants showed decreased optimum pH values in the acidic range (6.0-6.5). All of the variant L-AIs could be significantly activated by the addition of Co(2+) and Mn(2+). D478N and D478Q showed higher catalytic efficiencies (k cat/K m) toward D-galactose than that of wild-type L-AI. In addition, the D478N and D478Q variants exhibited a much higher conversion ratio of D-galactose to D-tagatose at 6.0 than the wild-type L-AI. According to the molecular model, residue D478 was located on the surface of the enzyme and distant from the active site. It was supposed that the charged state of residue 478 may influence the optimum pH for substrate binding or isomerization.