EMAIL THIS PAGE TO A FRIEND
Food microbiology
Quantitative detection of Toxoplasma gondii in tissues of experimentally infected turkeys and in retail turkey products by magnetic-capture PCR.
PMID 26338112
Abstract
Magnetic-capture PCR was applied for the quantitative detection of Toxoplasma gondii in tissues of experimentally infected turkeys and retail turkey meat products. For experimental infection, three T.xa0gondii strains (ME49, CZ-Tiger, NED), varying infectious doses in different matrices (organisms in single mouse brains or 10(3), 10(5), or 10(6) oocysts in buffer) were used. From all animals, breast, thigh, and drumstick muscle tissues and for CZ-Tiger-infected animals additionally brains and hearts were analyzed. Using the magnetic-capture PCR large volumes of up to 100xa0g were examined. Our results show that most T.xa0gondii parasites are present in brain and heart tissue. Of the three skeletal muscle types, drumsticks were affected at the highest and breast at the lowest level. Type III strain (NED) seems to be less efficient in infecting turkeys compared to type II strains, because only few tissues of NED infected animals contained T.xa0gondii DNA. Furthermore, the number of detected parasitic stages increased with the level of infectious dose. Infection mode by either oocyst or tissue cyst stage did not have an effect on the amount of T.xa0gondii present in tissues. In retail turkey meat products T.xa0gondii DNA was not detectable although a contact with the parasite was inferred by serology.