EMAIL THIS PAGE TO A FRIEND
International journal of hygiene and environmental health
Non-phthalate plasticizers in German daycare centers and human biomonitoring of DINCH metabolites in children attending the centers (LUPE 3).
PMID 26338253
Abstract
Plasticizers have been widely used for decades as additives in diverse applications, including consumer and building products, toys, cables, and floorings. Due to toxicological concerns and restrictions of different dialkyl ortho-phthalates, other plasticizers have been increasingly used in recent years. Therefore, di-isononyl cyclohexane-1,2-dicarboxylate (DINCH), di(2-ethylhexyl) terephthalate (DEHT), di(2-ethylhexyl) adipate (DEHA), acetyl tri-n-butyl citrate (ATBC), and trioctyl trimellitate (TOTM) plasticizer levels in indoor air and dust samples from 63 daycare centers in Germany were measured. Moreover, the urine samples of 208 children who attend 27 of these facilities were analyzed for the presence of four DINCH metabolites. DINCH, DEHT, and DEHA were present in indoor air with median values of 108 ng/m(3), 20 ng/m(3), and 34 ng/m(3), respectively. Median values of 302 mg/kg for DINCH, 49 mg/kg for DEHA, 40 mg/kg for DEHT, and 24 mg/kg ATBC were found in dust. In the urine samples, the three secondary metabolites of DINCH were observed with median values (95th percentiles) of 1.7 μg/l (10.0 μg/l) for OH-MINCH, 1.5 μg/l (8.0 μg/l) for oxo-MINCH, and 1.1 μg/l (6.1 μg/l) for cx-MINCH. Overall, these metabolite levels are orders of magnitude lower than the current HBM I values set by the German Human Biomonitoring Commission. Using general exposure assumptions, the intake resulting from dust ingestion and inhalation is low for children. The total daily DINCH intake calculated from biomonitoring data was 0.5 μg/kg b.w. using median values and 9.8 μg/kg b.w. as the maximum value. At present, non-phthalate plasticizers, especially DINCH, can be found in considerable amounts in dust samples from daycare centers and as DINCH metabolites in the urine of children. In relation to previous studies, the concentrations of DINCH in dust and urine have an increasing time trend. Compared with tolerable daily intake values, the total daily intake of DINCH reached only 1% of its maximum value to date; however, due to its increased use, higher exposure of DINCH is expected in the future.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
92124
Tris(2-ethylhexyl) trimellitate, Selectophore
C33H54O6