EMAIL THIS PAGE TO A FRIEND
Forensic science international
Optimization of spermatozoa detection using immunofluorescent staining and laser micro-dissection.
PMID 26338669
Abstract
The present study evaluated the use of an immunofluorescence-based assay for the microscopic detection of human spermatozoa, following which the fluorescence-labelled spermatozoa could be excised with a laser micro-dissection system. The Sperm Hy-Liter™ PI kit was able to detect spermatozoa from as little as 20nL of semen. No interference or non-specificity were observed when the kit was used on semen mixed with various body fluids such as blood and urine, as well as when semen was spiked onto different types of fabric. Good results could also be obtained with rectal samples which contain auto-fluorescent fecal materials through the use of dual FITC/PI filters. We also developed a method for concurrent testing of two protein biomarkers of semen (semenogelin and prostate-specific antigen) and detection of spermatozoa. This approach would maximize the evidential value from a single piece of sexual assault exhibit. The results also showed that staining by Sperm Hy-Liter™ PI does not interfere with DNA recovery, facilitating the generation of clear male DNA profiles from dissected spermatozoa, thereby making profile interpretation less complex. In summary, Sperm Hy-Liter™ PI staining was demonstrated to be sensitive, robust and specific.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
82804
3-(1-Pyridinio)-1-propanesulfonate, ≥97.0% (N)
C8H11NO3S