EMAIL THIS PAGE TO A FRIEND
Journal of microbiological methods
Enhancing recombinant protein solubility with ubiquitin-like small archeal modifying protein fusion partners.
PMID 26341610
Abstract
A variety of protein expression tags with different biochemical properties has been used to enhance the yield and solubility of recombinant proteins. Ubiquitin, SUMO (small ubiquitin-like modifier) and prokaryotic ubiquitin like MoaD (molybdopterin synthase, small subunit) fusion tags are getting more popular because of their small size. In this paper we report on the use of ubiquitin-like small archaeal modifier proteins (SAMPs) as fusion tags since they proved to increase expression yield, stability and solubility in our experiments. Equally important, they did not co-purify with proteins of the expression host and there was information that their specific JAB1/MPN/Mov34 metalloenzyme (JAMM) protease can recognize the C-terminal VSGG sequence when SAMPs fused, either branched or linearly to target proteins, and cleave it specifically. SAMPs and JAMM proteases from Haloferax volcanii, Thermoplasma acidophilum, Methanococcoides burtonii and Nitrosopumilus maritimus were selected, cloned, expressed heterologously in Escherichia coli and tested as fusion tags and cleaving proteases, respectively. Investigated SAMPs enhanced protein expression and solubility on a wide scale. T. acidophilum SAMPs Ta0895 and Ta01019 were the best performing tags and their effect was comparable to the widely used maltose binding protein (MBP) and N utilization substance protein A (NusA) tags. Moreover, H. volcanii SAMP Hvo_2619 contribution was mediocre, whereas M. burtonii Mbur_1415 could not be expressed. Out of four investigated JAMM proteases, only Hvo_2505 could cleave fusion tags. Interestingly, it was found active not only on its own partner substrate Hvo_2619, but it also cleaved off Ta0895.