EMAIL THIS PAGE TO A FRIEND
Virology
Trim21 regulates Nmi-IFI35 complex-mediated inhibition of innate antiviral response.
PMID 26342464
Abstract
In this study, using an immunoprecipitation coupled with mass spectrometry approach, we have identified the E3 ubiquitin ligase Trim21 as an interacting partner of IFI35 and Nmi. We found that this interaction leads to K63-linked ubiquitination on K22 residue of Nmi, but not IFI35. Using domain deletion analysis, we found that the interaction is mediated via the coiled-coil domain of Nmi and the carboxyl-terminal SPRY domain of Trim21. Furthermore, we show that depletion of Trim21 leads to significantly reduced interaction of Nmi with IFI35, which results in the abrogation of the negative regulatory function of the Nmi-IFI35 complex on innate antiviral signaling. Thus, Trim21 appears to be a critical regulator of the functions of the Nmi-IFI35 complex. Overall, the results presented here uncover a new mechanism of regulation of the Nmi-IFI35 complex by Trim21, which may have implications for various autoimmune diseases associated uncontrolled antiviral signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1406780
Anti-NMI antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1412622
ANTI-NMI antibody produced in mouse, clone 9E8, purified immunoglobulin, buffered aqueous solution
SAB1412636
ANTI-NMI antibody produced in mouse, clone 10E5, purified immunoglobulin, buffered aqueous solution
HPA008588
Anti-NMI antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV35736
Anti-NMI antibody produced in rabbit, IgG fraction of antiserum
SAB2101609
Anti-NMI antibody produced in rabbit, affinity isolated antibody
AV38248
Anti-TRIM21 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2501628
Anti-TRIM21 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406479
Anti-TRIM21 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA005673
Anti-TRIM21 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102542
Anti-TRIM21 antibody produced in rabbit, affinity isolated antibody
EHU045371 MISSION® esiRNA, esiRNA human TRIM21 (esiRNA1)
EMU072291 MISSION® esiRNA, esiRNA targeting mouse Trim21 (esiRNA1)
WH0009111M1
Monoclonal Anti-NMI antibody produced in mouse, clone 9D8, purified immunoglobulin, buffered aqueous solution
SAB1404672
Monoclonal Anti-NMI, (N-terminal) antibody produced in mouse, clone 9B8, purified immunoglobulin, buffered aqueous solution
SAB1402363
Monoclonal Anti-TRIM21 antibody produced in mouse, clone 2C9, purified immunoglobulin, buffered aqueous solution