EMAIL THIS PAGE TO A FRIEND
Gastroenterology
CSF1 Restores Innate Immunity After Liver Injury in Mice andxa0Serum Levels Indicate Outcomes of Patients With Acutexa0Liver Failure.
PMID 26344055
Abstract
Liver regeneration requires functional liver macrophages, which provide an immune barrier that is compromised after liver injury. The numbers of liver macrophages are controlled by macrophage colony-stimulating factor (CSF1). We examined the prognostic significance of the serum level of CSF1 in patients with acute liver injury and studied its effects in mice. Wexa0measured levels of CSF1 in serum samples collected from 55 patients who underwent partial hepatectomy at the Royal Infirmary Edinburgh between December 2012 and October 2013, as well as from 78 patients with acetaminophen-induced acute liver failure admitted to the Royal Infirmary Edinburgh or the University of Kansas Medical Centre. Wexa0studied the effects of increased levels of CSF1 in uninjured mice that express wild-type CSF1 receptor or a constitutive orxa0inducible CSF1-receptor reporter, as well as in chemokine receptor 2 (Ccr2)-/- mice; we performed fate-tracing experiments using bone marrow chimeras. We administered CSF1-Fc (fragment, crystallizable) to mice after partial hepatectomy and acetaminophen intoxication, and measured regenerative parameters and innate immunity by clearance of fluorescent microbeads and bacterial particles. Serum levels of CSF1 increased in patients undergoing liverxa0surgery in proportion to the extent of liver resected. Inxa0patients with acetaminophen-induced acute liverxa0failure, a low serum level of CSF1 was associated withxa0increased mortality. In mice, administration of CSF1-Fc promoted hepatic macrophage accumulation via proliferation of resident macrophages and recruitment of monocytes. CSF1-Fc also promoted transdifferentiation of infiltrating monocytes into cells with axa0hepatic macrophage phenotype. CSF1-Fc increased innatexa0immunity in mice after partial hepatectomy or acetaminophen-induced injury, with resident hepatic macrophage as the main effector cells. Serum CSF1 appears to be a prognostic marker for patients with acute liver injury. CSF1 might be developed as a therapeutic agent to restore innate immune function after liver injury.