EMAIL THIS PAGE TO A FRIEND
Scientific reports
Proteomic approaches for profiling negative fertility markers in inferior boar spermatozoa.
PMID 26348888
Abstract
The ability to predict male fertility is of paramount importance for animal breeding industries and for human reproduction. Conventional semen analysis generally provides information on the quantitative parameters of spermatozoa, but yields no information concerning its functional competence. Proteomics have identified candidates for male fertility biomarkers, but no studies have clearly identified the relationship between the proteome and sperm fertility. Therefore, we performed a proteomic analysis to investigate small and large litter size boar spermatozoa and identify proteins related to male fertility. In this study, 20 proteins showed differential expression levels in small and large litter size groups. Nineteen of these proteins exhibited decreased expression in large litter size samples and increased expression in the small litter group. Interestingly, only one protein was highly expressed in the large litter size spermatozoa. We then identified signaling pathways associated with the differentially expressed protein markers. Glutathione S-transferase Mu3 and glutathione peroxidase 4 were related to the glutathione metabolic pathway and arginine vasopressin receptor 2 was linked to vasopressin R2/STAT. In summary, this is the first study to consider negative fertility biomarkers, and the identified proteins could potentially be used as biomarkers for the detection of inferior male fertility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA046678
Anti-AVPR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502319
Anti-AVPR2 antibody produced in rabbit, affinity isolated antibody
SAB2900527
Anti-AVPR2 antibody produced in rabbit, affinity isolated antibody
SAB1405900
Anti-GSTM3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV53561
Anti-GSTM3 antibody produced in rabbit, affinity isolated antibody
SAB2103526
Anti-GSTM3 antibody produced in rabbit, affinity isolated antibody
HPA035190
Anti-GSTM3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
V5514 Anti-Vasopressin V2 Receptor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
U0631
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
CH4N2O
U5378
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
CH4N2O
U4883
Urea, 8 M (after reconstitution with 16 mL high purity water)
CH4N2O
U6504
Urea, for electrophoresis
CH4N2O
U4884
Urea, meets USP testing specifications
CH4N2O
U4128
Urea, BioReagent, suitable for cell culture
CH4N2O
U5128
Urea, ACS reagent, 99.0-100.5%
CH4N2O
U1250
Urea, ReagentPlus®, ≥99.5%, pellets
CH4N2O
51456
Urea, BioUltra, for molecular biology, ≥99.5% (T)
CH4N2O
33247
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
CH4N2O
15604
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
CH4N2O
U7129
Urea solution, 40 % (w/v) in H2O
CH4N2O
51457
Urea solution, BioUltra, ~8 M in H2O
CH4N2O
299367
Urea-12C, 99.9 atom % 12C
12CH4N2O
608734
Urea-14N2, 99.99 atom % 14N, 15N-depleted
CH414N2O