EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Annexin A7 modulates BAG4 and BAG4-binding proteins in mitochondrial apoptosis.
PMID 26349959
Abstract
Annexin A7 caused an anti-apoptosis phenotype in mouse hepatocarcinoma cells. Yet, the underlying mechanism remains unclear. In this study, we found that Annexin A7 had an inhibitory effect on the mitochondrial Cytochrome C release and Caspase-3 cleavage. Furthermore, Annexin A7 was identified to form a complex with BAG4 which is a negative regulator of apoptosis. Moreover, Annexin A7 was positively correlated with the protein levels of BAG4, Hsp70 and Bcl-2. Both Hsp70 and Bcl-2 bind BAG4 and inhibit Cytochrom C dependent activation of Caspase-3. Taken together, our study indicated Annexin A7 modulated BAG4 and BAG4-binding proteins in mitochondrial apoptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4475
Anti-Annexin VII antibody, Mouse monoclonal, clone 203-217/6, purified from hybridoma cell culture
GW22213A Anti-ANXA7 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV36584
Anti-ANXA7 (AB1) antibody produced in rabbit, IgG fraction of antiserum
GW22213B Anti-ANXA7 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV36585
Anti-ANXA7 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1405462
Anti-ANXA7 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2501206
Anti-BAG4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1400353
Anti-BAG4 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA018951
Anti-BAG4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405552
Anti-CAMK4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA011753
Anti-CAMK4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA017206
Anti-CAMK4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV37025
Anti-CAMK4 antibody produced in rabbit, IgG fraction of antiserum
SAB4503233
Anti-CAMK4 antibody produced in rabbit, affinity isolated antibody
SAB3500450
Anti-SODD antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV49121
Anti-ST6GALNAC4 antibody produced in rabbit, affinity isolated antibody
SAB2701445
Anti-ST6GALNAC4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1402474
Monoclonal Anti-BAG4 antibody produced in mouse, clone 2G9, purified immunoglobulin, buffered aqueous solution
WH0000814M1
Monoclonal Anti-CAMK4 antibody produced in mouse, clone 1A3, purified immunoglobulin, buffered aqueous solution