EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Expression of leukemia inhibitory factor in the rat retina following acute ocular hypertension.
PMID 26352383
Abstract
The aim of the present study was to investigate the expression of leukemia inhibitory factor (LIF) and its downstream signaling pathways in the rat retina following acute ocular hypertension. The intraocular pressure of the rats was elevated to 110 mmHg for 1 h by infusing the anterior chamber with normal saline. The retinal tissues were obtained 12 h, 24 h, and 2, 3 and 7 days after termination of the ocular hypertension. Hematoxylin and eosin and terminal deoxynucleotidyl transferase dUTP nick end labeling (TUNEL) staining were performed to assess the morphological changes and the apoptosis of retinal cells, respectively. Quantification of the retinal ganglion cells (RGCs) was performed using fluorogold retrograde (FG) staining. The expression levels of LIF, LIF receptor (LIFR), signal transducers and activators of transcription 3 (STAT3), phosphorylated STAT3 (P‑STAT3), Akt, phosphorylated‑Akt (P‑Akt), extracellular signal‑regulated kinase (ERK) and phosphorylated ERK (P‑ERK) were determined at different time‑points following acute ocular hypertension using western blot analysis. Reverse transcription‑quantitative polymerase chain reaction was performned to detect the mRNA expression levels of LIF and LIFR. The results revealed that 12 h, 24 h, 2, 3 and 7 days after reperfusion, the thickness of the inner nuclear layer and the inner plexiform layer was decreased, with a significant reduction in the number of RGCs, as determined using TUNEL and FG staining. The expression levels of LIF and LIFR were increased following acute ocular hypertension. At 12 h post‑retinal reperfusion, the expression levels of P‑STAT3 and P‑Akt were significantly upregulated, while the expression of P‑ERK was decreased. The changes in the expression levels of LIF and LIFR suggested that LIF may be important in the process of degeneration/protection following retinal ischemia induced by acute ocular hypertension, via activation of the Janus kinase/STAT and Akt signaling pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L9152 Anti-Leukemia Inhibitory Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
L9277 Anti-Leukemia Inhibitory Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
L1044 Anti-Leukemia Inhibitory Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
L1169 Anti-Leukemia Inhibitory Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB2502145
Anti-LIF antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406083
Anti-LIF antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA018844
Anti-LIF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101344
Anti-LIF antibody produced in rabbit, affinity isolated antibody
L0669 Monoclonal Anti-Leukemia Inhibitory Factor antibody produced in mouse, clone 9824, purified immunoglobulin, lyophilized powder
L0919 Monoclonal Anti-Leukemia Inhibitory Factor antibody produced in mouse, clone 9808, purified immunoglobulin, lyophilized powder
L0544 Monoclonal Anti-Leukemia Inhibitory Factor antibody produced in rat, clone 139124, purified immunoglobulin, lyophilized powder
SAB1402252
Monoclonal Anti-LIFR antibody produced in mouse, clone 4A10, purified immunoglobulin, buffered aqueous solution