PloS one
Corticotropin-Releasing Hormone Receptor Type 1 (CRHR1) Clustering with MAGUKs Is Mediated via Its C-Terminal PDZ Binding Motif.
PMID 26352593
Abstract
The corticotropin-releasing hormone receptor type 1 (CRHR1) plays an important role in orchestrating neuroendocrine, behavioral, and autonomic responses to stress. To identify molecules capable of directly modulating CRHR1 signaling, we performed a yeast-two-hybrid screen using the C-terminal intracellular tail of the receptor as bait. We identified several members of the membrane-associated guanylate kinase (MAGUK) family: postsynaptic density protein 95 (PSD95), synapse-associated protein 97 (SAP97), SAP102 and membrane associated guanylate kinase, WW and PDZ domain containing 2 (MAGI2). CRHR1 is co-expressed with the identified MAGUKs and with the additionally investigated PSD93 in neurons of the adult mouse brain and in primary hippocampal neurons, supporting the probability of a physiological interaction in vivo. The C-terminal PDZ (PSD-95, discs large, zona occludens 1) binding motif of CRHR1 is essential for its physical interaction with MAGUKs, as revealed by the CRHR1-STAVA mutant, which harbors a functionally impaired PDZ binding motif. The imitation of a phosphorylation at Thr413 within the PDZ binding motif also disrupted the interaction with MAGUKs. In contrast, distinct PDZ domains within the identified MAGUKs are involved in the interactions. Expression of CRHR1 in primary neurons demonstrated its localization throughout the neuronal plasma membrane, including the excitatory post synapse, where the receptor co-localized with PSD95 and SAP97. The co-expression of CRHR1 and respective interacting MAGUKs in HEK293 cells resulted in a clustered subcellular co-localization which required an intact PDZ binding motif. In conclusion, our study characterized the PDZ binding motif-mediated interaction of CRHR1 with multiple MAGUKs, which directly affects receptor function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21184P Anti-c-Myc (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW21222 Anti-c-Myc (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1306111
ANTI-CRFR1 (Q103) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900152
Anti-CRHR1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500465
Anti-CRHR1 antibody produced in rabbit, affinity isolated antibody
SAB2500319
Anti-DLG1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501245
Anti-DLG4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405717
Anti-DLG4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA010122
Anti-DLG4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401115
Anti-DLG4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2105773
Anti-DLG4 antibody produced in rabbit, affinity isolated antibody
SAB2100955
Anti-GPHN (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100956
Anti-GPHN (ab2) antibody produced in rabbit, affinity isolated antibody
HPA003116
Anti-GPHN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA024694 Anti-GPHN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
M3696 Anti-MAP2 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA008273
Anti-MAP2 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA012828
Anti-MAP2 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300658
Anti-MAP2 antibody produced in rabbit, affinity isolated antibody
M9942
Anti-MAP2 antibody, Mouse Monoclonal, clone HM-2, purified from hybridoma cell culture
SAB4300319
Anti-MYC (Ab-358) antibody produced in rabbit, affinity isolated antibody
SAB4300320
Anti-MYC (Ab-373) antibody produced in rabbit, affinity isolated antibody
SAB4300318
Anti-MYC (Ab-58) antibody produced in rabbit, affinity isolated antibody
SAB2103448
Anti-MYC, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4501941
Anti-MYC, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503661
Anti-MYC, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503662
Anti-MYC, C-Terminal antibody produced in rabbit, affinity isolated antibody
L5501
L-Lysine, ≥98% (TLC)
C6H14N2O2
62840
L-Lysine, crystallized, ≥98.0% (NT)
C6H14N2O2
W384704
L-Lysine, ≥97%
C6H14N2O2