EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
ApoG2 inhibits the antiapoptotic protein, Mcl‑1, and induces mitochondria‑dependent apoptosis in human colorectal cancer cells.
PMID 26352605
Abstract
Colorectal cancer (CRC) is a worldwide malignancy of high incidence and mortality. At present, there is a lack of effective drugs against CRC. The B‑cell leukemia/lymphoma 2 (Bcl‑2) protein family members are considered to be closely associated with tumorigenesis and the chemoresistance of CRC. As a novel gossypol derivative targeting antiapoptotic proteins of the Bcl‑2 family, apogossypolone (ApoG2) exhibits antitumor properties in various cancer types, although its effects against CRC remain to be fully elucidated. In the present study, the cytotoxicity of ApoG2 in vitro on CRC cells was investigated, with the aim of elucidating the underlying mechanism. Using an MTT assay, ApoG2 was revealed to inhibit the growth of the HT29, SW480 and HCT116 CRC cell lines in a dose‑ and a time‑dependent manner. Hoechst staining revealed that ApoG2 induced CRC cell apoptosis, marked by morphological changes, including cell shrinkage and nuclear fragmentation. Flow cytometric analysis also detected a higher apoptotic ratio following treatment with ApoG2. The ratio was dependent upon the concentration of ApoG2, which the cells were exposed to, and the duration of the exposure. Western blot analysis and immunoprecipitation experiments revealed that ApoG2 treatment led to the downregulation of the protein expression of Mcl‑1, and the interruption of the binding of Mcl‑1 to the protein Bax. Furthermore, treatment with ApoG2 led to the release of cytochrome c into the cytoplasm and the activation of caspases 3 and 7. The present study revealed that ApoG2 inhibited the proliferation of the CRC cell lines through mitochondrial signaling pathway‑dependent apoptosis, which may be associated with the disruption of the function of the Mcl‑1 protein by ApoG2.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W503401
2,4-Dimethylthiazole, ≥95%, FG
C5H7NS