PloS one
Matrix Metalloproteinase-2 (MMP-2) Gene Deletion Enhances MMP-9 Activity, Impairs PARP-1 Degradation, and Exacerbates Hepatic Ischemia and Reperfusion Injury in Mice.
PMID 26355684
Abstract
Hepatic ischemia and reperfusion injury (IRI) is an inflammatory condition and a significant cause of morbidity and mortality after surgery. Matrix metalloproteinases (MMPs) have been widely implicated in the pathogenesis of inflammatory diseases. Among the different MMPs, gelatinases (MMP-2 and MMP-9) are within the most prominent MMPs detected during liver IRI. While the role of MMP-9 in liver damage has been fairly documented, direct evidence of the role for MMP-2 activity in hepatic IRI remains to be established. Due to the lack of suitable inhibitors to target individual MMPs in vivo, gene manipulation is as an essential tool to assess MMP direct contribution to liver injury. Hence, we used MMP-2-/- deficient mice and MMP-2+/+ wild-type littermates to examine the function of MMP-2 activity in hepatic IRI. MMP-2 expression was detected along the sinusoids of wild-type livers before and after surgery and in a small population of leukocytes post-IRI. Compared to MMP-2+/+ mice, MMP-2 null (MMP-2-/-) mice showed exacerbated liver damage at 6, 24, and 48 hours post-reperfusion, which was fatal in some cases. MMP-2 deficiency resulted in upregulation of MMP-9 activity, spontaneous leukocyte infiltration in naïve livers, and amplified MMP-9-dependent transmigration of leukocytes in vitro and after hepatic IRI. Moreover, complete loss of MMP-2 activity impaired the degradation of poly (ADP-ribose) polymerase (PARP-1) in extensively damaged livers post-reperfusion. However, the administration of a PARP-1 inhibitor to MMP-2 null mice restored liver preservation to almost comparable levels of MMP-2+/+ mice post-IRI. Deficient PARP-1 degradation in MMP-2-null sinusoidal endothelial cells correlated with their increased cytotoxicity, evaluated by the measurement of LDH efflux in the medium. In conclusion, our results show for the first time that MMP-2 gene deletion exacerbates liver IRI. Moreover, they offer new insights into the MMP-2 modulation of inflammatory responses, which could be relevant for the design of new pharmacological MMP-targeted agents to treat hepatic IRI.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB19167
Anti-MMP-2 Antibody, whole molecule, Detect MMP-2 using this Anti-MMP-2 Antibody, whole molecule validated for use in IH(P) & WB.
C5670
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
CaCl2
429759
Calcium chloride, anhydrous, beads, −10 mesh, ≥99.99% trace metals basis
CaCl2
499609
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
CaCl2
C8106
Calcium chloride, meets USP testing specifications
CaCl2 · 2H2O
449709
Calcium chloride, anhydrous, beads, −10 mesh, ≥99.9% trace metals basis
CaCl2
21115
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
CaCl2
S2889
Sodium acetate, anhydrous, for molecular biology, ≥99%
C2H3NaO2
S3272
Sodium acetate, for electrophoresis, ≥99%, crystalline
C2H3NaO2
S5636
Sodium acetate, powder, BioReagent, for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
C2H3NaO2
S1429
Sodium acetate, meets USP testing specifications, anhydrous
C2H3NaO2
229873
Sodium acetate, 99.995% trace metals basis
C2H3NaO2
S7545
Sodium acetate, BioXtra, ≥99.0%
C2H3NaO2
71183
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
C2H3NaO2
W302406
Sodium Acetate Anhydrous, >99%, FCC, FG
C2H3NaO2
S8388
Sodium acetate solution, 0.3 M
C2H3NaO2
71196
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
C2H3NaO2
S8032
Sodium azide, BioXtra
N3Na
438456
Sodium azide, ≥99.99% trace metals basis
N3Na
S2002
Sodium azide, ReagentPlus®, ≥99.5%
N3Na
71289
Sodium azide, BioUltra, ≥99.5% (T)
N3Na
71290
Sodium azide, purum p.a., ≥99.0% (T)
N3Na