EMAIL THIS PAGE TO A FRIEND
Journal of cell science
Dexamethasone-induced cellular tension requires a SGK1-stimulated Sec5-GEF-H1 interaction.
PMID 26359301
Abstract
Dexamethasone, a synthetic glucocorticoid, is often used to induce osteoblast commitment of mesenchymal stem cells (MSCs), and this process requires RhoA-dependent cellular tension. The underlying mechanism is unclear. In this study, we show that dexamethasone stimulates expression of fibronectin and integrin α5 (ITGA5), accompanied by an increase in the interaction of GEF-H1 (also known as ARHGEF2) with Sec5 (also known as EXOC2), a microtubule (MT)-regulated RhoA activator and a component of the exocyst, respectively. Disruption of this interaction abolishes dexamethasone-induced cellular tension and GEF-H1 targeting to focal adhesion sites at the cell periphery without affecting dexamethasone-induced levels of ITGA5 and fibronectin, and the extracellular deposition of fibronectin at adhesion sites is specifically inhibited. We demonstrate that dexamethasone stimulates the expression of serum-glucocorticoid-induced protein kinase 1 (SGK1), which is necessary and sufficient for the induction of the Sec5-GEF-H1 interaction. Given the function of SGK1 in suppressing MT growth, our data suggest that the induction of SGK1 through treatment with dexamethasone alters MT dynamics to increase Sec5-GEF-H1 interactions, which promote GEF-H1 targeting to adhesion sites. This mechanism is essential for the formation of fibronectin fibrils and their attachment to integrins at adhesion sites in order to generate cellular tension.
Related Materials