EMAIL THIS PAGE TO A FRIEND
Journal of biomedical optics
Intraoperative laser speckle contrast imaging improves the stability of rodent middle cerebral artery occlusion model.
PMID 26359813
Abstract
Rodent middle cerebral artery occlusion (MCAO) model is commonly used in stroke research. Creating a stable infarct volume has always been challenging for technicians due to the variances of animal anatomy and surgical operations. The depth of filament suture advancement strongly influences the infarct volume as well. We investigated the cerebral blood flow (CBF) changes in the affected cortex using laser speckle contrast imaging when advancing suture during MCAO surgery. The relative CBF drop area (CBF50, i.e., the percentage area with CBF less than 50% of the baseline) showed an increase from 20.9% to 69.1% when the insertion depth increased from 1.6 to 1.8 cm. Using the real-time CBF50 marker to guide suture insertion during the surgery, our animal experiments showed that intraoperative CBF-guided surgery could significantly improve the stability of MCAO with a more consistent infarct volume and less mortality.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8877
2,3,5-Triphenyltetrazolium chloride, ≥95%
C19H15ClN4