PloS one
Autophagy Protects from Trastuzumab-Induced Cytotoxicity in HER2 Overexpressing Breast Tumor Spheroids.
PMID 26360292
Abstract
Multicellular tumor spheroids represent a 3D in vitro model that mimics solid tumor essential properties including assembly and development of extracellular matrix and nutrient, oxygen and proliferation gradients. In the present study, we analyze the impact of 3D spatial organization of HER2-overexpressing breast cancer cells on the response to Trastuzumab. We cultured human mammary adenocarcinoma cell lines as spheroids with the hanging drop method and we observed a gradient of proliferating, quiescent, hypoxic, apoptotic and autophagic cells towards the inner core. This 3D organization decreased Trastuzumab sensitivity of HER2 over-expressing cells compared to monolayer cell cultures. We did not observe apoptosis induced by Trastuzumab but found cell arrest in G0/G1 phase. Moreover, the treatment downregulated the basal apoptosis only found in tumor spheroids, by eliciting protective autophagy. We were able to increase sensitivity to Trastuzumab by autophagy inhibition, thus exposing the interaction between apoptosis and autophagy. We confirmed this result by developing a resistant cell line that was more sensitive to autophagy inhibition than the parental BT474 cells. In summary, the development of Trastuzumab resistance relies on the balance between death and survival mechanisms, characteristic of 3D cell organization. We propose the use of spheroids to further improve the understanding of Trastuzumab antitumor activity and overcome resistance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21985 Anti-CDKN1B antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22609 Anti-CDKN1B antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405601
Anti-CDKN1B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100391
Anti-CDKN1B antibody produced in rabbit, affinity isolated antibody
SAB4300350
Anti-ERBB2 (Ab-1221/1222) antibody produced in rabbit, affinity isolated antibody
SAB4300351
Anti-ERBB2 (Ab-1248) antibody produced in rabbit, affinity isolated antibody
SAB4300349
Anti-ERBB2 (Ab-877) antibody produced in rabbit, affinity isolated antibody
GW22631 Anti-ERBB2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA001383
Anti-ERBB2 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500782
Anti-HER2 antibody produced in rabbit, affinity isolated antibody
SAB4500783
Anti-HER2 antibody produced in rabbit, affinity isolated antibody
SAB4500787
Anti-HER2 antibody produced in rabbit, affinity isolated antibody
SAB4500789
Anti-HER2 antibody produced in rabbit, affinity isolated antibody
SAB4500784
Anti-HER2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500785
Anti-HER2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500786
Anti-HER2, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500788
Anti-HER2, C-Terminal antibody produced in rabbit, affinity isolated antibody
L7543
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB4200361
Anti-LC3B antibody, Mouse monoclonal, clone LC3B-6, purified from hybridoma cell culture
SAB2700738
Anti-Map1lc3b antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3501065
Anti-MAP1LC3B antibody produced in rabbit, affinity isolated antibody
HPA000451
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001164
Anti-MKI67 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306621
ANTI-P27KIP1 (N-TERM S12) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300058
Anti-phospho-ERBB2 (pTyr1221/pTyr1222) antibody produced in rabbit, affinity isolated antibody
SAB4300061
Anti-phospho-ERBB2 (pTyr1248) antibody produced in rabbit, affinity isolated antibody
SAB4300057
Anti-phospho-ERBB2 (pTyr877) antibody produced in rabbit, affinity isolated antibody
SAB4504586
Anti-phospho-HER2 (pTyr1112) antibody produced in rabbit, affinity isolated antibody
SAB4504425
Anti-phospho-HER2 (pTyr1221/Tyr1222) antibody produced in rabbit, affinity isolated antibody
SAB4504427
Anti-phospho-HER2 (pTyr877) antibody produced in rabbit, affinity isolated antibody