PloS one
Inhibition of IGF-1-Mediated Cellular Migration and Invasion by Migracin A in Ovarian Clear Cell Carcinoma Cells.
PMID 26360832
Abstract
Previously we isolated migracin A from a Streptomyces culture filtrate as an inhibitor of cancer cell migration. In the present research, we found that migracin A inhibited migration and invasion of ovarian clear cell carcinoma ES-2 cells. In the course of our mechanistic study, migracin A was shown to enhance vasohibin-1 expression in an angiogenesis array. We also confirmed that it increased the mRNA expression of this protein. Moreover, overexpression of vasohibin-1 lowered the migration but not the invasion of ES-2 cells. Then, we looked for another target protein employing a motility array, and found that migracin A lowered the IGF-1 expression. Knockdown of IGF-1 by siRNA decreased the migration and invasion of ES-2 cells. Migracin A also decreased Akt phosphorylation involved in the downstream signaling. Crosstalk analysis indicated that overexpression of vasohibin-1 decreased the IGF-1 expression. On the other hand, it showed no direct anticancer activity in terms of the ES-2 growth in agar. Migracin A inhibited the migration and IGF-1 expression in not only ES-2 but also another ovarian clear cell carcinoma JHOC-5 cells. In addition, it also inhibited capillary tube formation of human umbilical vein endothelial cells. Since its cytotoxicity is very low, migracin A may be a candidate for an anti-metastasis agent not exhibiting prominent toxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4500797
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500799
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500800
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500802
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500803
Anti-AKT antibody produced in rabbit, affinity isolated antibody
SAB4500796
Anti-AKT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500798
Anti-AKT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500004
Anti-AKT, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300335
Anti-AKT1 (Ab-308) antibody produced in rabbit, affinity isolated antibody
SAB4300575
Anti-AKT1 (Ab-450) antibody produced in rabbit, affinity isolated antibody
SAB4300334
Anti-AKT1 (Ab-473) antibody produced in rabbit, affinity isolated antibody
GW22757 Anti-AKT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW22775 Anti-AKT1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501934
Anti-AKT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3500216
Anti-Akt1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA002891
Anti-AKT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV06008 Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
SAB4500007
Anti-AKT1 antibody produced in rabbit, affinity isolated antibody
SAB4503853
Anti-phospho-Akt (pSer124) antibody produced in rabbit, affinity isolated antibody
SAB4504016
Anti-phospho-Akt (pSer129) antibody produced in rabbit, affinity isolated antibody
SAB4504015
Anti-phospho-Akt (pSer246) antibody produced in rabbit, affinity isolated antibody
SAB4504331
Anti-phospho-Akt (pSer473) antibody produced in rabbit, affinity isolated antibody
SAB4504332
Anti-phospho-Akt (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4504017
Anti-phospho-Akt (pThr72) antibody produced in rabbit, affinity isolated antibody
SAB4504110
Anti-phospho-Akt (pTyr326) antibody produced in rabbit, affinity isolated antibody
SAB4504014
Anti-phospho-Akt (pTyr474) antibody produced in rabbit, affinity isolated antibody
SAB4300042
Anti-phospho-AKT1 (pSer473) antibody produced in rabbit, affinity isolated antibody
SAB4300043
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4504749
Anti-phospho-AKT1 (pThr308) antibody produced in rabbit, affinity isolated antibody
SAB4503852
Anti-phospho-Akt1 (pThr450) antibody produced in rabbit, affinity isolated antibody