EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Jubanines F-J, cyclopeptide alkaloids from the roots of Ziziphus jujuba.
PMID 26361730
Abstract
Five Ib-type cyclopeptide alkaloids, jubanines F-J (1-5), and three known compounds, nummularine B (6), daechuine-S3 (7), and mucronine K (8) were isolated from the roots of Ziziphus jujuba. Their structures were fully characterized by spectroscopic analyses in combination with chemical derivatization. Compounds 1-3, and 6 were evaluated for their antiviral activity against the porcine epidemic diarrhea virus (PEDV). Compounds 2, 3, and 6 showed potent inhibitory effects on PEDV replication.
Related Materials