EMAIL THIS PAGE TO A FRIEND
Cancer research
YAP promotes malignant progression of Lkb1-deficient lung adenocarcinoma through downstream regulation of survivin.
PMID 26363011
Abstract
The serine/threonine kinase LKB1 is a well-characterized tumor suppressor that governs diverse cellular processes, including growth, polarity, and metabolism. Somatic-inactivating mutations in LKB1 are observed in about 15% to 30% of non-small cell lung cancers (NSCLC). LKB1 inactivation confers lung adenocarcinomas (ADC) with malignant features that remain refractory to therapeutic intervention. YAP activation has been linked to LKB1 deficiency, but the role of YAP in lung ADC formation and progression is uncertain. In this study, we showed that ectopic expression of YAP in type II alveolar epithelial cells led to hyperplasia in mouse lungs. YAP overexpression in the Kras(G12D) lung cancer mouse model accelerated lung ADC progression. Conversely, YAP deletion dramatically delayed the progression of lung ADC in LKB1-deficient Kras(G12D) mice. Mechanistic studies identified the antiapoptotic oncoprotein survivin as the downstream mediator of YAP responsible for promoting malignant progression of LKB1-deficient lung ADC. Collectively, our findings identify YAP as an important contributor to lung cancer progression, rationalizing YAP inhibition in the context of LKB1 deficiency as a therapeutic strategy to treat lung ADC.