EMAIL THIS PAGE TO A FRIEND
Monatshefte fur chemie
2-Allylaminothiazole and 2-allylaminodihydrothiazole derivatives: synthesis, characterization, and evaluation of bioactivity.
PMID 26366014
Abstract
Some reactions of selected chlorooxoesters and haloesters with a 1-allylthiourea under various conditions have been performed. The reactions have been performed in methanol in alkaline and neutral environment. Condensation of 1-allylthiourea with chlorooxoesters has been further led via acetal as intermediate compound. As a result, the compounds containing thiazole and a 4,5-dihydrothiazole ring with a good yield have been obtained. The structures of the compounds were verified by (1)H NMR, (13)C NMR as well as X-ray diffraction analysis. Due to the potential biological activity of the synthesized compounds, the parameters of their bioavailability have been determined, and the probability of pharmacological action has been defined. All of the obtained compounds fulfilled the rule of five, which indicate their good absorption after oral intake. The probability of pharmacological action and potential targets calculated for the obtained compounds show that they can be potential drugs.