EMAIL THIS PAGE TO A FRIEND
BMC complementary and alternative medicine
Ameliorating effects of gypenosides on chronic stress-induced anxiety disorders in mice.
PMID 26370834
Abstract
Ethanol extract from Gynostemma pentaphyllum (GP) shows anti-stress and anxiolytic functions in mice, and also protects dopamine neurons in 6-hydroxydopamine-lesioned rat model of Parkinson's disease. In addition, gypenosides (the gypenoside-enriched components of GP, GPS) have a protective effect on 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine-induced mouse model of Parkinson's disease. In this study, the ameliorating effects of GPS on chronic stress-induced anxiety disorders in mice were investigated. Mice were orally treated with GPS (100 and 200 mg/kg) once a day for 10 days, followed by exposure to electric footshock (EF) stress (0.6 mA, 1 s every 5 s, 3 min). After the final administration of either GPS, water extract of GP (GP-WX) or ethanol extract of GP (GP-EX, positive control), the behavioral tests such as elevated plus-maze, marble burying and locomotor activity tests, and the biochemical parameters including dopamine, serotonin and corticosterone levels, and c-Fos expression were examined. Treatment with GPS (100 and 200 mg/kg) increased the number of open arm entries and the time spent on open arms in elevated plus-maze which were reduced by chronic EF stress. GPS (100 and 200 mg/kg) reduced the number of marbles buried which increased by chronic EF stress. In these states, the brain levels of dopamine and serotonin decreased by chronic EF stress and they were recovered by GPS. The serum levels of corticosterone increased by chronic EF stress were also reduced by GPS (100 and 200 mg/kg). Finally, chronic EF stress-induced c-Fos expression was markedly reduced by GPS (100 and 200 mg/kg) in the brain. GPS (100 and 200 mg/kg) also showed an equivalent efficacy on anxiolytic functions, as compared with GP-EX (50 mg/kg). However, GP-WX (50 mg/kg) showed a less effect on anxiety disorders than GP-EX (50 mg/kg) and GPS (100 and 200 mg/kg). These results suggest that GPS (100 and 200 mg/kg) has anxiolytic effects on chronic EF stress-induced anxiety disorders by modulating dopamine and serotonin neuronal activities, c-Fos expression and corticosterone levels. GPS may serve as a phytonutrient in chronic stress-induced anxiety disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21144 Anti-c-Fos antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
F7799
Anti-c-Fos antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
F137 Anti-c-Fos antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
HPA018531
Anti-C-FOS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1306144
ANTI-C-FOS(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2100832
Anti-FOS (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100833
Anti-FOS (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2502116
Anti-FOS antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV100639
Anti-FOS antibody produced in rabbit, IgG fraction of antiserum
SAB2104185
Anti-FOS antibody produced in rabbit, affinity isolated antibody
SAB4500995
Anti-FOS antibody produced in rabbit, affinity isolated antibody
SAB4500993
Anti-FOS, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500996
Anti-FOS, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB5300463
Monoclonal Anti-FOS antibody produced in mouse, clone 2G2, ascites fluid
SAB1402201
Monoclonal Anti-FOS antibody produced in mouse, clone 2D11, purified immunoglobulin, buffered aqueous solution