EMAIL THIS PAGE TO A FRIEND
European journal of human genetics : EJHG
The C175R mutation alters nuclear localization and transcriptional activity of the nephronophthisis NPHP7 gene product.
PMID 26374130
Abstract
Nephronophthisis (NPH) is a rare autosomal ciliopathy, but the leading cause for hereditary end-stage renal disease in children. Most NPH family members form large protein networks, which appear to participate in structural elements of the cilium and/or function to restrict access of molecules to the ciliary compartment. The zinc-finger protein GLIS2/NPHP7 represents an exception as it has been implicated in transcriptional regulation; only two families with GLIS2/NPHP7 mutations and typical NPH manifestations have been identified so far. We describe here that the recently identified GLIS2/NPHP7(C175R) point mutation abolished the nuclear localization of GLIS2/NPHP7. Forced nuclear import did not rescue the transcriptional defects of GLIS2/NPHP7(C175R), indicating additional defects as DNA-binding protein. We further observed that wild type, but not GLIS2/NPHP7(C175R), prevented the cyst formation caused by depletion of nphp7 in zebrafish embryos. Taken together, our findings indicate that the C175R mutation affects both localization and function of GLIS2/NPHP7, supporting a role of this mutation in NPH, but questioning the direct involvement of GLIS2/NPHP7 in ciliary functions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3532
Phenol Red, powder, BioReagent, suitable for cell culture
C19H14O5S