EMAIL THIS PAGE TO A FRIEND
Developmental cell
Secrets of the Hair Follicle: Now on Your iPhone.
PMID 26374762
Abstract
Skin development requires communication between epithelial and mesenchymal cells, melanocytes, and neurons. In this issue of Developmental Cell, Sennett et al. (2015) shed new light on these mechanisms by simultaneously profiling multiple different cell types in embryonic mouse skin at the onset of hair follicle formation.