EMAIL THIS PAGE TO A FRIEND
Nanoscale
Quantum dot multiplexing for the profiling of cellular receptors.
PMID 26377627
Abstract
The profiling of cellular heterogeneity has wide-reaching importance for our understanding of how cells function and react to their environments in healthy and diseased states. Our ability to interpret and model cell behavior has been limited by the difficulties of measuring cell differences, for example, comparing tumor and non-tumor cells, particularly at the individual cell level. This demonstrates a clear need for a generalizable approach to profile fluorophore sites on cells or molecular assemblies on beads. Here, a multiplex immunoassay for simultaneous detection of five different angiogenic markers was developed. We targeted angiogenic receptors in the vascular endothelial growth factor family (VEGFR1, VEGFR2 and VEGFR3) and Neuropilin (NRP) family (NRP1 and NRP2), using multicolor quantum dots (Qdots). Copper-free click based chemistry was used to conjugate the monoclonal antibodies with 525, 565, 605, 655 and 705 nm CdSe/ZnS Qdots. We tested and performed colocalization analysis of our nanoprobes using the Pearson correlation coefficient statistical analysis. Human umbilical vein endothelial cells (HUVEC) were tested. The ability to easily monitor the molecular indicators of angiogenesis that are a precursor to cancer in a fast and cost effective system is an important step towards personalized nanomedicine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405819
Anti-FLT1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA011740
Anti-FLT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
SAB1408577
Anti-FLT1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
AV46700
Anti-NELL2 antibody produced in rabbit, IgG fraction of antiserum
HPA035715
Anti-NELL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21538 Anti-NRP2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA039980 Anti-NRP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411571
Anti-NRP2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
V1139 Anti-Vascular Endothelial Growth Factor Receptor 1 antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4500957
Anti-VEGFR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500958
Anti-VEGFR1, C-Terminal antibody produced in rabbit, affinity isolated antibody
AMAB90703
Monoclonal Anti-FLT1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0344, purified immunoglobulin, buffered aqueous glycerol solution
SAB1402857
Monoclonal Anti-NELL2 antibody produced in mouse, clone 1F6, purified immunoglobulin, buffered aqueous solution
SAB1409358
Monoclonal Anti-NELL2 antibody produced in mouse, clone 2D11, purified immunoglobulin, buffered aqueous solution
N3412 Monoclonal Anti-Neuropilin-2 antibody produced in mouse, clone 96009, purified immunoglobulin, lyophilized powder
WH0008828M1
Monoclonal Anti-NRP2 antibody produced in mouse, clone 3B8, purified immunoglobulin, buffered aqueous solution
V4762 Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-1 antibody produced in mouse, clone FLT-19, tissue culture supernatant
V4262
Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-1 antibody produced in mouse, clone FLT-11, ascites fluid