EMAIL THIS PAGE TO A FRIEND
Diagnostic pathology
Overexpression of Trps1 contributes to tumor angiogenesis and poor prognosis of human osteosarcoma.
PMID 26377811
Abstract
Trichorhinophalangeal syndrome 1 (Trps1) gene is a member of GATA transcription factor family and has an important function in tumorigenesis and progression. However, there are rare studies on its roles in carcinogenesis and prognostic significance in human osteosarcoma. The expression of Trps1 was detected by immunohistochemistry, and MVD was evaluated to determine the amounts of microvessels by counting CD31-positive endothelial cells. Of the 74 cases that underwent study, Trps1-positive cases were 24. And it was associated with MVD significantly (P = 0.008). The data also exhibited more cases of remote metastasis (P = 0.013) and higher Enneking stage (P = 0.017) in Trps1-positive group compared to Trps1-negative group. Univariate analysis revealed that distant metastasis, MVD and Trps1 expression were associated with a lower 3-year overall survival rate and disease-free survival rate (P = 0.003, and P = 0.012 respectively). Furthermore, Trps1 and distant metastasis retained their significant prognostic effects on patients survival rate by multivariate analysis (P < 0.05). Trps1 plays a crucial role in osteosarcoma angiogenesis, metastasis and clinical surgical stage. Trps1 can be a novel promising prognostic marker and therapeutic target, and antiangiogenic therapy which targets Trps1 molecule in patients with osteosarcoma may lead to improved prognosis and longer-term survival.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300902
Anti-TRPS1 antibody produced in rabbit, affinity isolated antibody