EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
Charge heterogeneity study of a Fc-fusion protein, abatacept, using two-dimensional gel electrophoresis.
PMID 26380522
Abstract
Medicinal products obtained by recombinant DNA technology are complex molecules and demonstrate a high degree of molecular heterogeneity. Charge heterogeneity and isoform pattern of this class of medicines, are parameters important for their quality, safety, and efficacy. In this study we report the application of two-dimensional gel electrophoresis (2-D electrophoresis) for the quality assessment, identification, charge heterogeneity and isoform pattern study of recombinant protein, CTLA4-Ig (abatacept), which has been selected as an example of the drug class, known as Fc-fusion proteins. In order to achieve an efficient separation of this complex analyte,2-D electrophoresis was optimized employing different experimental conditions regarding the selection of an immobilized pH gradient (IPG), sample pretreatment, presentation and detection procedure. Experimental datadocumented that 2-D electrophoresis is a suitable method for the assessment of identity, purity, structural integrity, isoform pattern and to monitor charge heterogeneity and post-translational glycosylation of the Fc-fusion protein, abatacept.