EMAIL THIS PAGE TO A FRIEND
Genome announcements
Genome Sequence of Rough and Smooth Variants of Pleomorphic Strain Lactobacillus farciminis CNCM-I-3699.
PMID 26383668
Abstract
The probiotic Lactobacillus farciminis CNCM-I-3699 is a pleomorphic strain exhibiting smooth and rough variants. We report their complete genomes consisting of a chromosome of 2, 4xa0Mb and a plasmid of 6,417xa0bp. The smooth variant differs by the presence of an additional plasmid of 35,418xa0bp.