EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
IRS2 and PTEN are key molecules in controlling insulin sensitivity in podocytes.
PMID 26384875
Abstract
Insulin signaling to the glomerular podocyte is important for normal kidney function and is implicated in the pathogenesis of diabetic nephropathy (DN). This study determined the role of the insulin receptor substrate 2 (IRS2) in this system. Conditionally immortalized murine podocytes were generated from wild-type (WT) and insulin receptor substrate 2-deficient mice (Irs2(-/-)). Insulin signaling, glucose transport, cellular motility and cytoskeleton rearrangement were then analyzed. Within the glomerulus IRS2 is enriched in the podocyte and is preferentially phosphorylated by insulin in comparison to IRS1. Irs2(-/-) podocytes are significantly insulin resistant in respect to AKT signaling, insulin-stimulated GLUT4-mediated glucose uptake, filamentous actin (F-actin) cytoskeleton remodeling and cell motility. Mechanistically, we discovered that Irs2 deficiency causes insulin resistance through up-regulation of the phosphatase and tensin homolog (PTEN). Importantly, suppressing PTEN in Irs2(-/-) podocytes rescued insulin sensitivity. In conclusion, this study has identified for the first time IRS2 as a critical molecule for sensitizing the podocyte to insulin actions through its ability to modulate PTEN expression. This finding reveals two potential molecular targets in the podocyte for modulating insulin sensitivity and treating DN.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2900230
Anti-INSR (AB1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900231
Anti-INSR (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1300890
Anti-INSR antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB2104752
Anti-INSR antibody produced in rabbit, affinity isolated antibody
HPA036302
Anti-INSR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106549
Anti-INSR antibody produced in rabbit, affinity isolated antibody
I7153 Anti-Insulin Receptor Substrate 1 (IRS-1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501555
Anti-IR antibody produced in rabbit, affinity isolated antibody
SAB4501556
Anti-IR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501557
Anti-IR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501558
Anti-IR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB3500281
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501568
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501569
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501570
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501571
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501572
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501573
Anti-IRS-1 antibody produced in rabbit, affinity isolated antibody
SAB4501567
Anti-IRS-1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300482
Anti-IRS1 (Ab-636) antibody produced in rabbit, affinity isolated antibody
SAB4300483
Anti-IRS1 (Ab-639) antibody produced in rabbit, affinity isolated antibody
SAB1406045
Anti-IRS1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1307241
ANTI-IRS2(C-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306059 ANTI-PTP1B (N-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1306060 ANTI-PTP1B antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
GW21488 Anti-PTPN1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1406342
Anti-PTPN1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA012542
Anti-PTPN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV45360
Anti-PTPN1 antibody produced in rabbit, affinity isolated antibody
SAB1411155
Anti-PTPN1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution