EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Selection of specific interactors from phage display library based on sea lamprey variable lymphocyte receptor sequences.
PMID 26391289
Abstract
Variable lymphocyte receptors (VLRs) are non-immunoglobulin components of adaptive immunity in jawless vertebrates. These proteins composed of leucine-rich repeat modules offer some advantages over antibodies in target binding and therefore are attractive candidates for biotechnological applications. In this paper we report the design and characterization of a phage display library based on a previously proposed dVLR scaffold containing six LRR modules [Wezner-Ptasinska et al., 2011]. Our library was designed based on a consensus approach in which the randomization scheme reflects the frequencies of amino acids naturally occurring in respective positions responsible for antigen recognition. We demonstrate general applicability of the scaffold by selecting dVLRs specific for lysozyme and S100A7 protein with KD values in the micromolar range. The dVLR library could be used as a convenient alternative to antibodies for effective isolation of high affinity binders.