EMAIL THIS PAGE TO A FRIEND
Experimental neurology
Thrombin decreases expression of the glutamate transporter GLAST and inhibits glutamate uptake in primary cortical astrocytes via the Rho kinase pathway.
PMID 26391563
Abstract
Astrocyte glutamate transporters GLAST and GLT1 play a key role in regulating neuronal excitation and their levels are altered in patients with epilepsy, and after traumatic brain injury. The mechanisms which regulate their expression are not well understood. We tested the hypothesis that exposure of astrocytes to high levels of thrombin, as may occur after a compromise of the blood-brain barrier, would reduce astrocyte glutamate transporter levels. In isolated rat cortical astrocytes we examined the effects of thrombin on the expression and function of glutamate transporters, and the signaling pathways involved in these responses by using Western blotting and selective inhibitors. Thrombin induced a selective decrease in the expression of GLAST but not GLT1, with a corresponding decrease in the capacity of astrocytes to take up glutamate. Activation of the thrombin receptor PAR-1 with an activating peptide induced a similar decrease in the expression of GLAST and compromise of glutamate uptake. The downregulation of GLAST induced by thrombin was mediated by the mitogen activated protein kinases p38 MAPK, ERK and JNK, but inhibition of these kinases did not prevent the decrease in glutamate uptake induced by thrombin. In contrast, inhibition of the Rho kinase pathway using the specific inhibitor, Y27632, suppressed both the decrease in the expression of GLAST and the decrease in glutamate uptake induced by thrombin. In hippocampal astrocyte cultures, thrombin caused a decrease in both GLAST and GLT1. In tissue resected from brains of children with intractable epilepsy, we found a decrease in the integrity of the blood-brain barrier along with a reduction in immunoreactivity for both transporters which was associated with an increase in cleaved thrombin and reactive astrogliosis. The in vitro results suggest a specific mechanism by which thrombin may lead to a compromise of astrocyte function and enhanced synaptic excitability after the blood-brain barrier is compromised. The human in vivo results provide indirect support evidence linking the compromise of the blood-brain barrier to thrombin-induced reduction in glutamate transporter expression and an increase in neuronal excitation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405777
Anti-F2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100739
Anti-F2 antibody produced in rabbit, affinity isolated antibody
HPA051476
Anti-F2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA054698
Anti-F2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106564
Anti-F2 antibody produced in rabbit, affinity isolated antibody
SAB2501233
Anti-F2R antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405778
Anti-F2R antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100740
Anti-F2R antibody produced in rabbit, affinity isolated antibody
SAB4501461
Anti-HSP90A, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2101093
Anti-HSP90AA1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2900334
Anti-HSP90AA1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2107614
Anti-HSP90AA1 antibody produced in rabbit, affinity isolated antibody
SAB2107615
Anti-HSP90AA1 antibody produced in rabbit, affinity isolated antibody
SAB4300541
Anti-HSP90AB1 (Ab-254) antibody produced in rabbit, affinity isolated antibody
SAB2701832
Anti-hsp90ab1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701806
Anti-hsp90ab1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV09016
Anti-HSP90AB1 antibody produced in rabbit, IgG fraction of antiserum
SAB2107344
Anti-HSP90AB1 antibody produced in rabbit, affinity isolated antibody
SAB4501462
Anti-HSP90B antibody produced in rabbit, affinity isolated antibody
SAB4501463
Anti-HSP90B antibody produced in rabbit, affinity isolated antibody
AV09018 Anti-HSPCB antibody produced in rabbit, IgG fraction of antiserum
SAB1305467
ANTI-HSPCB(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
GW21241 Anti-HSPN antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA038790
Anti-MARK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500760
Anti-MARK2 antibody produced in rabbit, affinity isolated antibody
GW20092F Anti-Prothrombin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
T5700 Anti-Thrombin Receptor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
WH0002147M1
Monoclonal Anti-F2 antibody produced in mouse, clone 1B7, purified immunoglobulin, buffered aqueous solution
SAB1402186
Monoclonal Anti-F2R antibody produced in mouse, clone 2C5, purified immunoglobulin, buffered aqueous solution
SAB5300449
Monoclonal Anti-HSP90AA1 antibody produced in mouse, clone 5G5, purified immunoglobulin, buffered aqueous solution