EMAIL THIS PAGE TO A FRIEND
Autophagy
Common γ-chain cytokine signaling is required for macroautophagy induction during CD4+ T-cell activation.
PMID 26391567
Abstract
Macroautophagy is a cellular process that mediates degradation in the lysosome of cytoplasmic components including proteins and organelles. Previous studies have shown that macroautophagy is induced in activated T cells to regulate organelle homeostasis and the cell's energy metabolism. However, the signaling pathways that initiate and regulate activation-induced macroautophagy in T cells have not been identified. Here, we show that activation-induced macroautophagy in T cells depends on signaling from common γ-chain cytokines. Consequently, inhibition of signaling through JAK3, induced downstream of cytokine receptors containing the common γ-chain, prevents full induction of macroautophagy in activated T cells. Moreover, we found that common γ-chain cytokines are not only required for macroautophagy upregulation during T cell activation but can themselves induce macroautophagy. Our data also show that macroautophagy induction in T cells is associated with an increase of LC3 expression that is mediated by a post-transcriptional mechanism. Overall, our findings unveiled a new role for common γ-chain cytokines as a molecular link between autophagy induction and T-cell activation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PRS3611
Anti-Beclin-1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3613
Anti-Beclin-1 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2103299
Anti-BECN1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1407194
Anti-DNMBP antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA037878
Anti-DNMBP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410413
Anti-DNMBP antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2101189
Anti-JAK3 antibody produced in rabbit, affinity isolated antibody
SAB4501603
Anti-JAK3 antibody produced in rabbit, affinity isolated antibody
SAB4300133
Anti-phospho-STAT1 (pSer727) antibody produced in rabbit, affinity isolated antibody
SAB4504538
Anti-phospho-STAT1 (pSer727) antibody produced in rabbit, affinity isolated antibody
S2565 Anti-phospho-STAT1 (pTyr701) antibody produced in rabbit, affinity isolated antibody, aqueous glycerol solution
SAB4300032
Anti-phospho-STAT1 (pTyr701) antibody produced in rabbit, affinity isolated antibody
SAB4504539
Anti-phospho-STAT1 (pTyr701) antibody produced in rabbit, affinity isolated antibody
SAB4300039
Anti-phospho-STAT6 (pThr645) antibody produced in rabbit, affinity isolated antibody
SAB4504545
Anti-phospho-STAT6 (pThr645) antibody produced in rabbit, affinity isolated antibody
SAB4300038
Anti-phospho-STAT6 (pTyr641) antibody produced in rabbit, affinity isolated antibody
S5061 Anti-phospho-STAT6 (pTyr641) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504546
Anti-phospho-STAT6 (pTyr641) antibody produced in rabbit, affinity isolated antibody
SAB1406748
Anti-SQSTM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB3500430
Anti-SQSTM1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104334
Anti-SQSTM1 antibody produced in rabbit, affinity isolated antibody
SAB1105169 Anti-STAT1 (671-685) antibody produced in rabbit, IgG fraction of antiserum
SAB4300326
Anti-STAT1 (Ab-701) antibody produced in rabbit, affinity isolated antibody
SAB4300441
Anti-STAT1 (Ab-727) antibody produced in rabbit, affinity isolated antibody
GW22758 Anti-STAT1 (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV33373
Anti-STAT1 (AB1) antibody produced in rabbit, affinity isolated antibody
GW22780 Anti-STAT1 (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
AV33374
Anti-STAT1 (AB2) antibody produced in rabbit, affinity isolated antibody
AV38933
Anti-STAT1 (AB3) antibody produced in rabbit, affinity isolated antibody
AV38934
Anti-STAT1 (AB4) antibody produced in rabbit, affinity isolated antibody